VIDEO: See How Sinter Feed Gets Blended and Analyzed
sinter videoIn this animated video, you’ll see how bulk materials, including sinter, get blended and analyzed during online elemental analysis. Sinter is used to convert iron into steel; and sinter plants, in combination with blast furnaces, are also used in non-ferrous smelting. The animation will take you through the process of sinter feed analysis and blending (raw mix feed, additives, mixing drum, ignition hood, sinter strand), and then elemental and basicity analysis, bulk material chemical analysis, input to process control system, additive control to improve sinter chemistry and quality and decrease return fines.
Watch the 6-minute Sinter Feed Blending and Analysis Video.