Infographic: All About Silver
silver mineralAccording to the United States Geological Survey Minerals Report, last year U.S. mines produced approximately 1,100 tons of silver with an estimated value of $570 million. There was an additional reported production of commercial-grade silver with an estimated total output of 2,100 tons from domestic and foreign ores and concentrates and from old and new scrap.
Silver has been known since ancient times, and yet is currently a major component of the newest electronics.  In addition, it can be found in jewelry, coins, photographic materials, medical equipment, and even as an ingredient in plastics.
To ensure silver is pure, or identify other metals that may be alloyed with it, portable XRF analyzers for precious metals can provide a fast, accurate, and most importantly, a nondestructive method to test the purity and composition of all precious metals, including silver. XRF quickly determines the presence and concentration of other trace alloying elements including lead, a toxic metal.
Learn more about this highly malleable, reflective, and ductile metal in our latest Infographic: 9 Fast Facts About Silver..
Infographic about silver
Download the Infographic: 9 Fast Facts About Silver.
Chris Calam
Written by:
Chris Calam
Senior Sales Manager, Thermo Fisher Scientific
Chris Calam spent over 26 years at Thermo Fisher Scientific, focused on optical emission, X-Ray fluorescence and diffraction, X-ray for electron microscopes across the UK, in the industrial, research and academia markets.
Read more Calam, Chris