  • Contact
Accelerating ScienceAdvancing Mining / Coal / Working in the Coal Mine – Avoiding Dust Inhalation
Working in the Coal Mine – Avoiding Dust Inhalation
Written by Alan Matta | Published: 12.09.2014
coal dustPrevious posts in this series walked you through what it’s like to actually work in a coal mine. Some of the conditions aren’t that pleasant, and are possibly even dangerous. In this post we’ll focus on one of those potential dangers—dust—and what is being done about it.
Dust inhalation is a huge concern in coal, metal, nonmetal, stone, and sand and gravel mining operations. In fact, the U.S. Mine Safety and Health Administration (MSHA) considers respirable coal dust to be one of the most serious occupational hazards in the mining industry. Coal and crystalline silica dust are the causes of serious, sometimes fatal lung diseases such as pneumoconiosis (CWP, or black lung), which affects coal miners, as well as silicosis, tuberculosis, chronic bronchitis, emphysema, chronic obstructive pulmonary disease (COPD), and chronic renal disease, which affect coal and other types of miners. The National Institute for Occupational Safety and Health (NIOSH) has classified crystalline silica as a potential occupational carcinogen.
CWP is on the Rise
According to the NIOSH Office of Mine Safety and Health Research (OMSHR) web site, CWP is on the rise in certain areas of the U.S., following a long decline. A nationwide pneumoconiosis surveillance program administered by the NIOSH Division of Respiratory Disease Studies (DRDS) found that 2.6 % of underground coal mine workers examined during 2005-2009 had some degree of CWP. A newly published report shows that 2% of examined surface coal mine workers had some degree of CWP even though most of them had never worked in underground mines. Guidelines for the implementation of dust reducing technologies are available through the NIOSH website.
Organizations such as NIOSH and MSHA are continually developing programs to identify risks, monitor dust levels, and implement strategies and technologies to reduce exposure. These are important steps in reducing the overall dust level at the site, and use of additional tools will help workers assess their exposure and encourage actions to help minimize risks. A significant advancement in this area is the emergence of the continuous personal dust monitor (CPDM).
MSHA has specified that a CPDM will be used for compliance dust sampling in the new Dust Rule, which was released into the Federal Register on May 1, 2014, and will require use of a CPDM in underground coal mines as of February 1, 2016. The original CPDM  real-time dust monitor was mutually developed by NIOSH and Thermo Fisher Scientific in a collaboration that began over 16 years ago using Thermo Scientific patented technologies.
Automated Personal Dust Monitoring is Equal to or Better than the Current Manual Dust Collection
Extensive NIOSH testing in laboratory and underground mine tests with differing coal dust composition and moisture content has demonstrated that this technology enables accurate dust sampling and meets all the requirements defined under 30 CFR part 74. The results indicate that the automated personal dust monitoring is equal to or better than the current manual dust collection and analysis method used since 1972 (personal monitoring program began in 1970).
CPDM data is available immediately after the end of a miner’s shift and shorter term dust exposure data is available continuously. This provides the miner with their current and projected exposure, permitting the miner to make adjustments in their work routine to avoid high concentrations of dust. The CPDM provides real time mass measurement through use of a TEOM (tapered element oscillating microbalance).  This technology provides true mass measurement rather that a nephelometer, which are by nature as very precise, but not necessarily accurate as they use the size of the particle and assumes a specific weight for each particle.
The CPDM also incorporates a cyclone rather than an impactor for more precise cut point of of 4.5 microns, which NIOSH has determined is the most effective cut point for measuring respirable coal dust. The CPDM also has a heater element to drive off excessive moisture and a momentum compensator, which eliminates frequency changes attributed to movement.   Fugitive Dust is another coal mine concern.  
The term fugitive dust refers to particulate that is lifted into the air either by man-made or natural activities in large open areas.  See our previously published fugitive dust article.  
Share this article
5
SHARES
FacebookLinkedinTwitterMail
Antimony (Sb) mine, mining. Stibnite ore (antimonite, antimony trisulfide) background. Critical mineral, raw material. Metal used solder, bullets, plain bearings, alloys, lead-acid batteries
INFOGRAPHIC: 9 Fast Facts About Antimony
It’s a silvery, lustrous grey and a semi-metallic toxic el... Marlene Gasdia-Cochraneby / 07.15.2025
Read More
air surrounding cement factory
What Is Ambient Air?
Learn about ambient air, its significance, pollutants, and m... by Marlene Gasdia-Cochrane / 06.06.2025
Read More
Safety helmets and gloves hang from a rack on a mining site
Happy Miners Day!
The US Department of Labor Mine Safety and Health Administra... by Marlene Gasdia-Cochrane / 12.06.2024
Read More
Shield design bundle element vector idea with creative modern style
What are the Largest Diamonds and How Are they Found?
In previous articles, we wrote about the January 1905 discov... Marlene Gasdia-Cochraneby / 08.23.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Comments
  1. nitonxrf says
    December 18, 2014 at 4:56 pm
    We received a comment on this post via a tweet:
    “While miners’ R in an ‘enclosed’ environment, inhaling fumes created from the heat of a welder’s tool is of respiratory concern.”
    Here is the author’s response:
    Thank you for your comment. The great majority of fumes from welding operations results are very small in particle size (0.25 – 1.50 microns), but some can exceed 10 microns. The PDM3600 (http://www.thermoscientific.com/en/product/pdm3600-personal-dust-monitor.html) will include those particles smaller than 4.5 microns as part of the total concentration reported. The instrument measures all particles smaller than 4.5 microns (respirable fraction – this size enters the lungs), with the majority in underground coal mines operation being coal dust.
    However, that total concentration can include particles from non coal dust sources. Examples of non coal courses include silica in areas of high rock content, rock dust from dust suppression efforts, diesel fumes from equipment as well as sources such as welding operations.
    It may be possible to use a filter collection system, such as the CMDPSU, to collect samples in the area where the welding operations are being conducted to collect a representative sample of the particle in that area and perform a post monitoring analysis to identify the source of the specific particulate. This is the collection method used to help identify silica content (post monitoring analysis is usually performed using X-ray diffraction to isolate the silica).
    Please let me know if you need additional information.
    Reply
  2. card-course.com says
    August 26, 2020 at 9:40 pm
    I’d like to find out more? I’d like to find out somke additional information.
    Reply
    • Marlene Gasdia-Cochrane, Editor says
      September 22, 2020 at 3:17 pm
      Learn about dust monitors here: https://www.thermofisher.com/order/catalog/product/PDM3700#/PDM3700
      Reply
  3. all CBSA codes of LA says
    August 30, 2020 at 4:41 am
    I read this post fully about the resemblance of most recent and preceding technologies, it’s awesome
    article.
    Reply
  4. fit yourself says
    August 30, 2020 at 7:16 pm
    This iinfo is worth everyone’s attention. How can I find out more?
    Reply
    • Marlene Gasdia-Cochrane, Editor says
      September 22, 2020 at 3:08 pm
      Subscribe to our Advancing Mining blog and receive educational articles every week!
      Reply
  5. https://Who-Isit.com/ says
    September 12, 2020 at 8:56 pm
    Thank you for some other gret post. Where else may just anybody
    get that type of info in such a perfect method of writing?
    I’ve a presentation next week, and I am at the search for such information.
    Reply
  6. protect-yourdata.com says
    September 28, 2020 at 5:08 am
    My brother recommended I might like this blog.
    He was entirely right.This post actually
    mace my day. You cann’timagine ust how much time I had spent for this info!
    Thanks!
    Reply
  7. laphonezip.com says
    November 5, 2020 at 6:48 am
    Right here is thee right site for anyone who wjshes to fibd out aabout this topic.
    You realize so much its almost tough to argue with you (not that I really
    would want to…HaHa). You certainly put a new spin on a topic
    that’s been written about for decades. Wonderful stuff, just wonderful!
    Reply
  8. Efficient CBSA dictionary in Los Angeles says
    November 5, 2020 at 10:06 am
    This wesite certainly has all the information I wanted concerning this subject and didn’t know who to ask.
    Reply
  9. https://usadataprotection.com/ says
    November 12, 2020 at 8:49 pm
    Hurrah! Finally I got a weblog from where
    I know how to really take useful data conerning my
    study and knowledge.
    Reply
  10. All Fips Codes Of Los Angeles says
    November 12, 2020 at 10:18 pm
    I don’t even know how I stopped up right here, but I
    believed this submit was great. I do not know who you’re however certainly you’re going to a
    famous blogger for those who are not already.
    Cheers!
    Reply
Trackbacks
  1. An End to Black Lung Disease: How Close Are We? (Part 1) says:
    June 15, 2016 at 4:23 pm
    […] Coal and crystalline silica dust cause other serious, sometimes fatal lung diseases such as silicosis, tuberculosis, chronic bronchitis, emphysema, chronic obstructive pulmonary disease (COPD), and chronic renal disease. The National Institute for Occupational Safety and Health (NIOSH) has classified crystalline silica as a potential occupational carcinogen. Read the previous post, Working in the Coal Mine – Avoiding Dust Inhalation. […]
    Reply
  2. The MSHA Dust Rule and How Mines Can Comply says:
    July 13, 2016 at 2:53 pm
    […] you’ve read our Working in the Coal Mine series, you’re familiar with the very serious issue of dust inhalation, a huge concern in coal, metal, nonmetal, stone, and sand and gravel mining […]
    Reply
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Analysis of Flowback Water for Reuse
Are You Ready for MATS? Here’s What you Need to Know
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook