Where Electron Microscopy Fits in the AI Ecosystem
Artificial intelligence is often discussed in terms of algorithms, models, and compute performance. Headlines focus on GPUs, large-scale training clusters, and new model capabilities. But beneath that visible layer lies a complex ecosystem, one that spans chip design, fabrication, packaging, and data storage.
What ultimately determines the pace of AI innovation is not just how fast we can design new architectures, but how effectively we can build, validate, and scale the chips that power them.
Electron microscopy (EM) plays a critical role in this ecosystem, spanning workflows across focused ion beam (FIB) milling, scanning electron microscopy (SEM), and transmission electron microscopy (TEM), while complementing related electrical fault isolation techniques. This technology enables the industry to see and understand structures at the nanoscale. In doing so, we help accelerate innovation across every layer of the AI hardware stack.
Understanding the AI hardware stack: A system of interdependencies
To understand where EM fits in, it is useful to map the AI hardware ecosystem into a few key areas:
  • Design—architecture and validation of AI chips
  • Fabless and foundry manufacturing—high-volume semiconductor production
  • Memory—DRAM, 3D NAND, and emerging memory technologies
  • Advanced packaging—chiplets, 2.5D/3D integration, high-bandwidth memory (HBM)
  • Data storage—long-term storage at data centers
AI chip linked to memory, advanced packaging, storage, design, and semiconductor manufacturing.
Figure 1. The interdependencies of the semiconductor AI hardware stack.
Each of these areas is pushing the limits of physics. Transistor dimensions are measured in nanometers, interconnects approach atomic scales, and packaging stacks become increasingly complex. At these dimensions, traditional inspection and analysis methods fall short.
This is where EM and failure analysis become indispensable.
AI chip design: Accelerating debug cycles with FIB milling
As AI accelerators grow in complexity, so does the need for rapid silicon debug and design iterations. Advanced AI chips often require post-silicon modifications to validate functionality, isolate design issues, and optimize performance without waiting for a new fabrication cycle.
EM tools play a critical role in enabling these workflows:
  • FIB milling supports advanced-node AI chip development by enabling sub-micron circuit edits (“nano-surgery”) with localized interconnect rerouting, reducing turnaround time for silicon debug, functional verification, and prototype optimization.
By enabling precise modifications directly on silicon, engineers can quickly test design changes, validate hypotheses, and determine whether observed behavior originates from the design itself or from manufacturing variation. This capability significantly accelerates root-cause analysis and shortens iteration cycles.
Circuit edit showing a conductive connection to an exposed device node for electrical debugging.
Figure 2. Circuit edit showing a deposited, conductive connection to an exposed device node, enabling signal rerouting for electrical debug.
Fabless and foundry manufacturing: Enabling yield at advanced nodes with EM
As leading-edge nodes approach 2 nm and beyond, semiconductor manufacturing requires near-atomic–scale precision. Small variations in materials or patterning can significantly impact yield.
EM supports critical steps in process development and control:
  • Nanoprobing places ultra-fine probes onto individual device nodes to localize a failing transistor in GAA devices
  • FIB-SEM prepares sub-15 nm thick TEM lamellae to isolate the unique transistor for further analysis in the TEM
  • TEM automates energy dispersive X-ray spectroscopy (EDS) to reveal residual SiGe resulting from incomplete channel-release etching
These tools help manufacturers identify issues such as voids, contamination, opens/shorts, and pattern fidelity challenges introduced by EUV lithography.
Yield directly affects the availability and cost of AI hardware. Without consistent, high-yield manufacturing, scaling AI infrastructure becomes limited.
TEM-EDS elemental map showing nitrogen, oxygen, silicon and germanium in a SiGe recess etch 20 nm scale
Figure 3. TEM EDS elemental mapping of SiGe recess etch.
Memory: Supporting AI’s data demands with EM tools
AI workloads are fundamentally data intensive. Whether training large models or running inference at scale, performance is often constrained by memory bandwidth and capacity rather than compute alone.
Technologies like 3D NAND and emerging 3D DRAM are central to addressing this demand, but they come with scaling challenges such as increasing layer counts in 3D NAND, maintaining cell integrity in 3D DRAM, and managing variability in densely packed structures.
EM tools enable detailed characterization of these systems:
  • Plasma FIB-SEM enables automated in-line metrology of 300-plus–layer 3D NAND devices by rapidly revealing buried channel structures, allowing engineers to quickly assess etching quality, bowing, twisting, and critical dimension (CD) variation, improving fab process control.
By helping improve understanding at the materials and structural level, EM helps drive improvements in both memory performance and reliability.
Argon plasma-FIB milling revealing through-volume NAND devices.
Figure 4. (Left) Argon plasma-FIB milling revealing through-volume NAND devices. (Center) SEM image with nanometer-scale measurements of NAND channel critical dimensions. (Right) Plot showing individual channel diameters versus height through the stack.
Advanced packaging: Performance meets complexity
AI performance is driven not just by individual chips but by how they are integrated. Advanced packaging technologies such as chiplets, 2.5D interposers, and 3D stacking enable higher bandwidth, lower latency, and improved energy efficiency.
But they also introduce new challenges, including dense interconnects and through-silicon vias (TSVs), thermal stress across heterogeneous materials, and interface defects that impact signal integrity.
EM provides the data needed to address these challenges:
  • Lock-in thermography identifies failure-induced hotspots in packaged devices through highly sensitive thermal mapping
  • Plasma FIB-SEM is then used to create a cross-section revealing the defect
As packaging becomes a primary driver of system performance, the ability to analyze and optimize these structures at nanoscale resolution becomes essential.
Plasma FIB cross-section showing through-silicon via (TSV) misalignment.
Figure 5. TSV misalignment exposed using plasma-FIB cross-section milling.
Data storage: Electron microscopy for HDD manufacturing
The rate of data generation is rapidly increasing, driven in part by AI applications. This data must be stored and accessed with high reliability. It is housed in data centers using distributed storage systems that combine solid-state drives (SSDs) and hard disk drives (HDDs).
EM tools have become essential for the design, ramp, and high-volume manufacturing of read/write heads in modern HDDs, especially with the introduction of HAMR (heat-assisted magnetic recording).
  • FIB-SEM provides in-line, automated, cross-sectional metrology of the write head
  • TEM delivers ultra-precise metrology of the read head, whose critical structures are substantially smaller and more sensitive to dimensional variation
The data storage industry continues to innovate, and EM tools are central to the development of new storage technologies.
We have a unique position across the ecosystem
What differentiates us in the AI landscape is the breadth of our engagement. We provide the critical instrumentation and analysis capabilities that enable every player in the ecosystem.
Our strength lies in integration, automation, and 40 years of expertise in the semiconductor industry. This combination allows us to operate across all layers of the hardware stack, from early design validation to high-volume manufacturing and field reliability.
As AI systems become more powerful and more pervasive, the need for this level of insight will only grow. We look forward to working with our customers in emerging areas like silicon photonics and helping the industry build the future of AI.
Sasan Ziatabar