Prepacked Chromatography Columns
On this page:
Selecting chromatography columns for biomolecule separation
Chromatography columns separate proteins, nucleic acids, and other biomolecules. Column selection depends on the stationary phase, operating pressure, and scale of use. Thermo Fisher Scientific supports accelerated speed to market, reproducibility and ease of use from development through manufacturing with prepacked columns.
Prepacked columns
Prepacked chromatography columns aim to simplify downstream workflows by reducing the need for manual packing. Available in multiple formats and volumes, these columns are packed with resins to facilitate purification across research, process development, and manufacturing. Options include RoboColumn™, GoPure, and MiniChrom formats, offering flexibility for evaluation and scale-up.
Choose the right chromatography column for your workflow
Chromatographic column design influences recovery and scalability, so matching the format to the workflow is crucial when transitioning from development to manufacturing. MiniChrom columns and small-scale GoPure formats are commonly used for resin evaluation and bench-scale development. The RoboColumn prepacked column format is intended for automated high-throughput optimization with robotic systems. Preparative prepacked columns are designed to extend these studies from bench-scale to pilot scale to clinical manufacturing.
GMP-scale prepacked chromatography columns
Thermo Scientific POROS GoPure chromatography columns bring together our innovative AAV affinity and polish resins—each conveniently prepacked in its own column—to deliver a complete, ready-to-use purification solution. Easily integrated into your AAV workflow, these columns can help you achieve high purity and performance with minimal set-up. 
Why choose our AAV prepacked chromatography columns?
  • Improved operational efficiency—Convenient, easy-to-use columns
  • Consistent performance—With capture and polish resins
  • Reduced lead times—Shipped in as little as 1–2 weeks
  • Documented quality—Packed in an IS07 environment
  • Streamlined AAV workflow—Minimize set-up time and operational complexities 
Affinity chromatography columns
Polish chromatography columns
Small-scale prepacked chromatography columns
GoPure small-scale prepacked columns
GoPure columns offer prepacked formats for resin screening and process development. Available in 1 mL and 5 mL sizes, they support process development with consistent bed heights for reliable scale-up.
MiniChrom columns
MiniChrom columns are prepacked formats designed to mimic process-scale columns. Offered in 1 mL and 5 mL sizes, they are used for method development and scale-down studies.
RoboColumn prepacked columns
RoboColumn formats are prepacked in 8-column strips for automated, parallel separations on robotic liquid handling systems. Available in 50 µL, 200 µL, and 600 µL volumes, these columns support high-throughput screening and process development.
Explore chromatography techniques
Chromatographic techniques—such as affinity, ion-exchange, hydrophobic interaction, and mixed-mode chromatography—are used at different stages of purification. Each mode applies distinct binding principles, making it important to match the technique with your target molecule and process needs.
Discover other chromatography products
Thermo Fisher Scientific offers additional chromatography products to optimize purification. Explore resins, membranes, and screening plates developed to complement column-based workflows and facilitate flexibility across scales.
CaptureSelect and POROS Technologies
CaptureSelect affinity resin leverages highly specific ligands for the capture of antibodies, proteins, viral vectors, and other biomolecules. POROS resin uses tuned chemistry on a rigid backbone with large throughpores for process-scale separations, offering high resolution and capacity.
Chromatography resins
Chromatography resins are available for affinity, ion exchange (IEX), hydrophobic interaction (HIC), and mixed-mode workflows. Formats span from evaluation tools to production-scale resins applied in cGMP biomanufacturing.
Membrane Chromatography
Membrane chromatography supports higher flow rates and shorter processing times compared to traditional packed-bed resin and can help accelerate biologics production from development through manufacturing.
96-Well Screening Plates
A 96-well plate format allows scientists to streamline resin evaluation by running multiple purifications in parallel. This enables early comparison of binding and wash conditions before moving to larger-scale studies.
Chromatography resources
Chromatography resources offer protocols, application notes, and videos that cover resin selection, column formats, and purification strategies. Content spans ion exchange, hydrophobic interaction, mixed-mode, and affinity chromatography, including Protein A chromatography.
RoboColumn™ is a registered trademark of Repligen Corporation.
For Research Use Only. Not for use in diagnostic procedures.