| Product | Activity assay | Binding assay | Pack size | Number of standard wells |
|---|
| Kinase | X | X | 100 μg | Variable |
| 5X Kinase Buffer A1,2 | X | X | 4 mL of 5X | 2,000 x 20 μL |
| Phosphospecific Antibody3 | X | | 25 μg 1 mg | 4,000 x 20 μL 160,000 x 20 μL
|
Substrate
| X
|
| 1 mg (peptide) 1 mL (poly-GT OR -GAT) 20 nmol (protein)
| 125,000 (except Fl-p53)5 16,700 x 20 μL 10,000 x 20 μL
|
| Antibody Dilution Buffer4 | X | | 100 mL | 10,000 x 20 μL |
| 500 mM EDTA | X | | 1 mL | 2,500 x 20 μL |
| 10 mM ATP | X | | 500 μL | Variable (e.g., 5,000 x 20 μL at 100 μM ATP) |
| Kinase Tracer6 | | X | 25 μL in 100% DMSO | Variable (e.g., 830 x 15 μL at 100 nM Tracer 236) |
| Anti-epitope Tag Antibody3 | | X | 25 μg 1 mg | 5,300 x 15 μL 210,000 x 15 μL |
| Control Inhibitor | X | X | Kinase-specific (often staurosporine,7 PHZ1271 100 μg) | N/A |
- Kinase Buffer A is supplied as a 5X concentrated stock. Prepare a 1X solution by adding 4 mL of the 5X solution to 16 mL of ultrapure water. The 1X Kinase Buffer A is stable at room temperature. 1X Kinase Buffer A consists of 50 mM HEPES, pH 7.5, 10 mM MgCl2, 1 mM EGTA, and 0.01% Brij-35.
- Some kinases require additives such as calmodulin and Ca2+, which need to be added to the Kinase Buffer A. Specific additives can be found in individual Kinase LanthaScreen® Activity validation packets.
- Prior to use, the antibody tube should be centrifuged at approximately 10,000 x g for 10 minutes, and the volume needed for the assay should be carefully pipetted from the supernatant.
- The antibody dilution buffer does not contain EDTA. EDTA is added separately to TR-FRET dilution buffer, followed by the addition of antibody.
- Assays containing Fl-p53 as a substrate are run at a much higher concentration of substrate, so 1 mg will run 29,500 wells.
- Check the tube for the concentration of tracer. For Tracer 236, this is 50 μM in DMSO; for Tracer 178, 199, 314, or 1710, this is 25 μM in DMSO.
- A 1 mM stock of staurosporine can be prepared by dissolving 100 μg of staurosporine in 210 μL of DMSO. The molecular weight of staurosporine is 466.53 Da.
Note: Kinase reaction buffer is the general buffer name used to indicate that Kinase Buffer A may also contain additives.
Note: Antibody centrifugation is required to remove aggregates whose Tb or Eu donor signals can disrupt data analysis.