On this page:
Accurate identification the first time
Engineered to deliver accurate genotypic results in less than 5 hours, MicroSEQ PCR and Sequencing Reagent Kits facilitate a streamlined workflow from DNA amplification to sequencing. 
Each kit includes the necessary components, including proprietary master mixes and primers. They have been thoroughly tested and optimized for use with the SeqStudio family of genetic analyzers, helping ensure optimal performance and ease-of-use. Whether you are identifying bacterial or fungal isolates, the MicroSEQ PCR and Sequencing Kits provides high-resolution Sanger sequencing, enabling you to achieve reliable and reproducible results. 
MicroSEQ kits include:
PCR Kits
Sequencing Kits
2X Master Mix
Positive control DNA
Negative control DNA
3X Forward sequencing mix
3X Reverse sequencing mix
Bacterial identification
The target for bacterial identification is the 16S ribosomal RNA (rRNA) gene sequence. The 16S rRNA is ubiquitous and can therefore be used to study phylogenetic relationships among all bacteria. The gene is 1,500 base pairs (bp) long and contains 9 variable regions. The conserved regions are identical in all micro-organisms and can be used for amplification via PCR, while the divergent regions are used for identification through sequencing. 
With the MicroSEQ ID method, users have the option of choosing to sequence the first 500 bp of the 16S rDNA gene or the full 1,500 bp. Kits and the supporting validated library databases are available for both options. The first 500 bp are sufficient for a routine identification that encompasses 3 of the 9 hypervariable regions of the 16S gene (Figure 1). In some cases, the 500 bp region is not enough to discriminate among very closely related bacteria and therefore requires a more informative full gene read. Furthermore, sequencing of the entire 1,500 bp sequence is required when describing a new species.
Figure 1. Hypervariable regions within the 16S rRNA gene. There are 9 hypervariable regions within the bacterial 16S gene, indicated in green. The conserved regions are indicated in grey.
Bacterial ID kits and libraries
Description
PCR kit
Sequencing kit
Library
# of entries
16sR full gene
MicroSEQ Full Gene 16S rDNA PCR Kit
MicroSEQ Full Gene 16S rDNA Sequencing Kit
MicroSEQ ID 16S rDNA Full Gene Library v2.1
1,262
16sR, first 500 base pairs
Fast MicroSEQ 500 16S rDNA PCR Kit
MicroSEQ 500 16S rDNA Sequencing Kit
MicroSEQ ID 16S rDNA 500 Microbial Library, v2022
2,100
MicroSEQ ID 16S rDNA 500 Supplemental Library, v2022
7,645
Fungal and yeast identification
Comparative sequence analyses of both the expansion region D2 of the larger rRNA molecule in the large subunit of the eukaryotic ribosome (LSU-D2) and ITS regions have been successfully used for the identification and classification of fungi down to the species level. The ITS region exhibits more variability, which can give rise to many more “sequence types” than the D2 region when used for sequence-based fungal identification. Fungal genomes may contain more than 100 copies of the rDNA cluster; it therefore is critical to understand and differentiate between simple sequence variability and sequence variability that reflects actual relatedness. The MicroSEQ fungal identification database is derived from sequences from LSU-D2 rDNA, which helps deliver highly reliable fungal identification and classification in routine identification tests.
Fungal and yeast ID kits and libraries
Description
PCR kit
Sequencing kit
Library
# of entries
LSU-D2 rDNA
Fast MicroSEQ D2 LSU rDNA Fungal PCR Kit
MicroSEQ D2 rDNA Fungal Sequencing Kit
MicroSEQ ID Fungal Gene Library, v2022
1,737
Request a technical consultation
No matter your questions, we are here to help. Our experienced team of scientists and technical specialists is ready to assist you in choosing the right solution and products. Complete this form to start the conversation.
Analytics Knowledge Hub
Explore the Analytics Knowledge Hub and discover a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize analytical testing in your bioprocess workflow.
Additional bioprocessing resources
Discover areas where you can cut costs and gain efficiency using our integrated calculator.
Explore our collection of information and insights to support your drug development journey.
Learn, evaluate, connect.
Connect with a Bioprocessing representative or request more info.
For Research Use Only. Not for use in diagnostic procedures.