Product Image
Applied Biosystems™

Fast MicroSEQ™ 500 16S rDNA PCR Kit

The Fast MicroSEQ™ 500 16S rDNA PCR kit utilizes new Fast technology and provides bacterial identifications from colony to results in as little as 5 hours using the Veriti™ 96-well Thermal Cycler.
Have Questions?
Número de catálogoCantidad
43704891 kit
Número de catálogo 4370489
Precio (MXN)
25,104.64
Each
Cantidad:
1 kit
Precio (MXN)
25,104.64
Each

The Fast MicroSEQ™ 500 16S rDNA PCR kit utilizes new Fast technology and provides bacterial identifications from colony to results in as little as 5 hours using the Veriti™ 96-well Thermal Cycler. This kit is also compatible with the GeneAmp™ PCR System 9700. The PCR-amplified products that are generated using this kit can be sequenced using the MicroSEQ™ 500 16S rDNA Seqencing Kit. The two kits together permit the amplification and sequencing of 500 base pairs of the 16S rRNA gene. These sequences can then be compared to the MicroSEQ™ ID Analysis Software 16S rDNA 500 Library to obtain an identity match. This kit does not include a protocol or a Quick Reference Card.

  • Sequence-based measurement ensures you get reproducible results in under 24 hours.
  • Growth requirement-independent measurement enables you to identify bacteria using objective criteria.
  • Universal domain-level primers eliminate up-front optimization.
  • MicroSEQ™ ID Analysis Software ensures accurate species identification.
  • Customizable libraries give you the flexibility to create new libraries and search validated libraries that contain the most frequently encountered organisms.
  • Requires minimal training-same work flow as the MicroSEQ™ D2 LSU fungal kit.

The MicroSEQ™ ID System: A Complete, Integrated Solution
Streamlining every step in microbial identification, Applied Biosystems™ MicroSEQ™ System combines the advantages of MicroSEQ™ ID Software with easy PrepMan Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To ensure optimal performance and system-wide integration, we develop and test all components of the system together.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (equipo)Veriti Thermal Cycler
BibliotecasCustomizable, Bacterial Library (> 2000 species)
Línea de productosMicroSEQ
Tipo de productorDNA PCR Kit
Cantidad1 kit
Tipo de muestraADN (genómico)
Condiciones de envíoDry Ice
SoftwareMicroSEQ™ ID Analysis Software
Objetivo1st 500 bp of 16S rRNA gene
Clase de organismo dianaBacteria
TécnicaSBT (Sequence-Based Typing)
Método de detecciónSonda de cebado
Para utilizar con (aplicación)Standard PCR
GC-Rich PCR PerformanceLow
Velocidad de reacciónFast
Unit SizeEach
Contenido y almacenamiento
Each kit contains enough PCR Master Mix to perform 50 PCR amplifications, 5 negative control assays, and 5 positive control assays. One tube of positive control E. coli DNA, and one of water (for use as a negative control) are also included.

Store at -15°C to -25°C upon reciept. After first use, store at 2°C to 8°C in a PCR Clean Room.

Preguntas frecuentes

Do you have stability information for the FAST MicroSEQ 500 16S rDNA PCR Kit (Cat. No. 4370489) when subjected to temperature fluctuations?

The FAST MicroSEQ 500 16S rDNA PCR Kit is stable if stored at -20 degrees C for 15-16 months. We do not have any information available for temperatures outside of this range.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

Citations & References (159)

Citations & References
Abstract
Structures of aldouronic acids liberated from kenaf xylan by endoxylanases from Streptomyces sp.
Authors:Komiyama, H; Enomoto, A; Sueyoshi, Y; Nishio, T; Kato, A; Ishii, T; Shimizu, K
Journal:
PubMed ID:
'Streptomyces sp. SK519 was cultivated in the medium containing 0.5% birch wood xylan to produce extracellular xylanase. The xylanase was isolated from the culture supernatant by ammonium sulfate precipitation and partially purified by absorbing on the insoluble xylan. Alkali-extracted kenaf glucuronoylan of which main chain primarily consisting of (1-4)-linked β-D-xylopyranose ... More
Pseudomonas panipatensis sp. nov., isolated from an oil-contaminated site
Authors:Gupta, SK; Kumari, R; Prakash, O; Lal, R
Journal:
PubMed ID:
'A Gram-negative, motile, rod-shaped, non-sporulating, aerobic bacterial strain (Esp-1(T)) was isolated from oil-contaminated soil of Panipat Oil Refinery, India, and its taxonomic position was determined using a polyphasic approach. Strain Esp-1(T) grew in the presence of 2% NaCl at 30 °C and was characterized chemotaxonomically by having C(16:0) as the ... More
Multilocus DNA sequence comparisons rapidly identify pathogenic molds.
Authors:Rakeman JL; Bui U; Lafe K; Chen YC; Honeycutt RJ; Cookson BT
Journal:
PubMed ID:
'The increasing incidence of opportunistic fungal infections necessitates rapid and accurate identification of the associated fungi to facilitate optimal patient treatment. Traditional phenotype-based identification methods utilized in clinical laboratories rely on the production and recognition of reproductive structures, making identification difficult or impossible when these structures are not observed. We ... More
Species distribution and antimicrobial resistance of enterococci isolated from surface and ocean water
Authors:Moore, DF; Guzman, JA; Mcgee, C
Journal:
PubMed ID:
'Aims: The species identification and antimicrobial resistance profiles were determined for enterococci isolated from Southern California surface and ocean waters. Methods and Results: Species identification was determined for 1413 presumptive Enterococcus isolates from urban runoff, bay, ocean and sewage water samples. The most frequently isolated species were Enterococcus faecalis, Enterococcus ... More
Multicenter evaluation of the new VITEK 2 advanced colorimetric yeast identification card.
Authors:Hata DJ; Hall L; Fothergill AW; Larone DH; Wengenack NL
Journal:
PubMed ID:
'The performance of the new VITEK 2 Advanced Colorimetry yeast identification (YST) card for use with the VITEK 2 system (bioMerieux, Inc., Hazelwood, MO) was compared to that of the API 20C AUX (API) system (bioMerieux SA, Marcy-l''Etoile, France) in a multicenter evaluation. A total of 12 quality control, 64 ... More