Search
MTT is a colorimetric assay used to assess cell viability as a function of cell number based on metabolic activity. The water-soluble MTT is converted to an insoluble formazan, which is then solubilized and the concentration determined by optical density at 570 nm.
Jurkat cells were diluted to the indicated cell numbers in 100 µL volumes, delivered to the wells of a microplate, and incubated for 4 hours with the CyQUANT MTT Cell Viability Assay. Absorbance measurements at 570 nm were made using a microplate reader.
For Research Use Only. Not for use in diagnostic procedures.