    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • ELISA Kits ›
          • Neuroserpin ELISA Kits
          This product was previously sold as SKU 900-K412
          Invitrogen
          Human Neuroserpin ELISA Development Kit (ABTS), PeproTech®
          View all (2) Neuroserpin ELISA Kits
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          Technical guide & protocol Tech Support
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Human Neuroserpin ELISA Development Kit (ABTS)
          Group 53 Created with Sketch.
          Human Neuroserpin ELISA Development Kit (ABTS)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Human Neuroserpin ELISA Development Kit (ABTS)
          Human Neuroserpin ABTS ELISA standard curve (Product # 900-K412K). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Human Neuroserpin ELISA Development Kit (ABTS)
          Human Neuroserpin ELISA Development Kit (ABTS) (900-K412K)
          videoThumbnail
          ►
          Product Specifications
          Analytical sensitivity
          Lot-specific
          Assay range
          Lot-specific
          Hands-on time
          1 hr 45 min
          Time-to-result
          24 hr 30 min
          Homogenous (no wash)
          No
          Instrument
          Absorbance Microplate Reader
          Product size
          10 x 96 Tests
          Contents
          This Human Neuroserpin ELISA Kit includes Capture Antibody, Detection Antibody, Protein Standard, and HRP-conjugate. The HRP-conjugate is packaged separately.
          Storage
          Refer to product documentation for component specific storage temperature.
          Protein name
          Neuroserpin
          Protein family
          Growth Factors & Receptors
          Species (tested)
          Human
          Detector antibody conjugate
          Biotin
          Label or dye
          HRP
          About This Kit
          900-K412K consists of: 900-K412 (capture antibody, detection antibody, and protein standard), and AVIDIN-60UL (avidin HRP-conjugate).
          Human Neuroserpin ABTS ELISA Development Kit contains the key components required for the quantitative measurement of natural and/or recombinant human Neuroserpin in a sandwich ELISA format. Human Neuroserpin ABTS ELISA Development Kit contains enough material to assay the target cytokine in at least 1,000 ELISA plate wells.
          The ABTS ELISA Buffer Kit (Product # 900-K00), sold separately, is recommended to be used in conjunction with this ABTS ELISA Development Kit for optimal performance. This buffer kit contains all the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions.
          Target information
          This gene encodes a member of the serpin superfamily of serine proteinase inhibitors. The protein is primarily secreted by axons in the brain, and preferentially reacts with and inhibits tissue-type plasminogen activator. It is thought to play a role in the regulation of axonal growth and the development of synaptic plasticity. Mutations in this gene result in familial encephalopathy with neuroserpin inclusion bodies (FENIB), which is a dominantly inherited form of familial encephalopathy and epilepsy characterized by the accumulation of mutant neuroserpin polymers. Multiple alternatively spliced variants, encoding the same protein, have been identified.
          Bioinformatics
          Gene aliases : HNS-S1, HNS-S2, neuroserpin, PI12
          Gene ID : (Human) 5274
          Gene symbol : SERPINI1
          Protein Aliases : DKFZp781N13156, isofom of neuroserpin, isoform of human neuroserpin, novel alternatively spliced truncated isoform of human neuroserpin, Neuroserpin, peptidase inhibitor 12, PI-12, serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1, serpin I1, serpin peptidase inhibitor clade I (neuroserpin) member 1 isoform SERPINI1_1, serpin peptidase inhibitor clade I (neuroserpin) member 1 isoform SERPINI1_2, serpin peptidase inhibitor clade I member 1, serpin peptidase inhibitor, unnamed protein product
          UniProt ID (Human) Q99574
          Recommended Products
          IL-6 Human Matched Antibody Pair
          Because you are interested in IL-6 Human ELISA Kit.
          SIZE
          10 x 96 tests
          PRICE
          USD 400.00
          Cat # CHC1263
          guarantee_icon
          Performance Guarantee
          If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-7fb9f846c9-wx6nt:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0