Search
Thermo Scientific GeneRuler DNA Ladders contain mixtures of individual chromatography-purified DNA fragments, designed to deliver precision and ease-of-use for everyday electrophoresis.
The chromatography-purified DNA fragments in GeneRuler ladders produce high quality reference bands. In routine gel electrophoresis, GeneRuler DNA ladders produce clean, crisp bands in the gel with recognizable patterns (Figure 1).
In capillary electrophoresis, GeneRuler DNA ladders produces sharp peaks (Figure 2). The mass of the DNA fragments in GeneRuler ladders are verified spectrophotometrically; this allows for the estimation of the mass of sample DNA with greater accuracy.
Figure 1. A comparison of reference bands produced by GeneRuler DNA ladder and other commercially available DNA ladders. GeneRuler DNA ladders produce clean and crisp reference bands, with an easy-to-remember pattern. The volume of DNA ladders added to each lane was in accordance with manufacturer recommendation (A) Lane 1: GeneRuler 1 kb DNA Ladder, ready-to-use; lane 2: GeneRuler 1 kb Plus DNA Ladder, ready-to-use; lane 3: Sigma-Aldrich DirectLoad™ Wide Range DNA Marker; lane 4: NEB Quick-Load™ 2-Log DNA Ladder (0.1–10.0 kb); lane 5: NEB Quick-Load™ 1 kb DNA Ladder. (B) Lane 1: GeneRuler 100 bp DNA Ladder, ready-to-use; lane 2: GeneRuler 100 bp Plus DNA Ladder, ready-to-use; lane 3: Sigma-Aldrich DirectLoad™ PCR 100 bp Low Ladder; lane 4: NEB Quick-Load™ Purple 100 bp DNA Ladder.
Figure 2. A comparison of peak widths produced by GeneRuler DNA ladder and other commercially available DNA ladders. GeneRuler DNA ladder produces sharp peaks compared to broad and double peaks produced by competitor products. (A) Thermo Scientific GeneRuler 100 bp DNA Ladder (red) and Promega 100 bp DNA Ladder (blue). (B) GeneRuler 100 bp DNA Ladder (red) and Sigma-Aldrich DirectLoad™ PCR 100 bp Low Ladder (blue).
The TriTrack DNA Loading Buffer, 6X contains three dyes (bromophenol blue, xylene cyanol FF, and orange G) for visual tracking of DNA migration during electrophoresis. These dyes do not interfere with visualization.
Ready-to-use DNA ladders premixed with the TriTrack Loading Buffer are stable at room temperature up to six months.
GeneRuler DNA ladders are earth-friendly products. Organic solvents are not used in production of Thermo Scientific DNA ladders, which helps reduce environmental waste from manufacturing processes. Furthermore, GeneRuler ladders are stable at ambient temperature and shipped in non-insulated shipping boxes, which generate less waste compared to insulated boxes.
GeneRuler DNA ladders can be ideal for DNA sizing and approximate quantification of a wide range of double-stranded DNA fragments ranging between 10 bp and 48.5 kb.
GeneRuler DNA ladders are available in two formats:
GeneRuler DNA Ladders are routinely used in the molecular biology techniques such as DNA cloning, PCR, and reverse transcription. Choose the DNA ladder that best fits your needs.
| Product name | GeneRuler Ultra Low Range DNA Ladder | GeneRuler Low Range DNA Ladder | GeneRuler 50 bp DNA Ladder | GeneRuler 100 bp DNA Ladder | GeneRuler 100 bp Plus DNA Ladder |
|---|---|---|---|---|---|
| Ready-to-use format | SM1213 | SM1193 | SM0373 | SM0243 SM0244 | SM0323 SM0324 |
| Colorless DNA ladders (dye not premixed) | SM1211 SM1212 | SM1191 SM1192 | SM0371 SM0372 | SM0241 SM0242 | SM0321 SM0322 |
| Base pairs (bp) | 10–300 | 25–700 | 50–1,000 | 100–1,000 | 100–3,000 |
![]() | ![]() | ![]() | ![]() | ![]() |
* Bands represent each of the DNA fragments. The dark bands represent reference bands.
| Product name | GeneRuler 1 kb DNA Ladder | GeneRuler 1 kb Plus DNA Ladder | GeneRuler Express DNA Ladder | GeneRuler DNA Ladder Mix | GeneRuler High Range DNA Ladder |
|---|---|---|---|---|---|
| Ready-to-use format | SM0314 SM0313 | SM1333 SM1334 | SM1553 | SM0334 SM0333 | SM1353 |
| Colorless DNA ladders (dye not premixed) | SM0311 SM0312 | SM1331 SM1332 | SM1551 SM1552 | SM0331 SM0332 | SM1351 SM1352 |
| Base pairs (bp) | 250–10,000 | 75–20,000 | 100–5,000 | 100–10,000 | |
![]() | ![]() | ![]() | ![]() | ![]() |
* Bands represent each of the DNA fragments. The dark bands represent reference bands.
| Product name | GeneRuler Ultra Low Range DNA Ladder | GeneRuler Low Range DNA Ladder | GeneRuler 50 bp DNA Ladder | GeneRuler 100 bp DNA Ladder | GeneRuler 100 bp Plus DNA Ladder |
|---|---|---|---|---|---|
| Ready-to-use format | SM1213 | SM1193 | SM0373 | SM0243 SM0244 | SM0323 SM0324 |
| Colorless DNA ladders (dye not premixed) | SM1211 SM1212 | SM1191 SM1192 | SM0371 SM0372 | SM0241 SM0242 | SM0321 SM0322 |
| Base pairs (bp) | 10–300 | 25–700 | 50–1,000 | 100–1,000 | 100–3,000 |
![]() | ![]() | ![]() | ![]() | ![]() |
* Bands represent each of the DNA fragments. The dark bands represent reference bands.
| Product name | GeneRuler 1 kb DNA Ladder | GeneRuler 1 kb Plus DNA Ladder | GeneRuler Express DNA Ladder | GeneRuler DNA Ladder Mix | GeneRuler High Range DNA Ladder |
|---|---|---|---|---|---|
| Ready-to-use format | SM0314 SM0313 | SM1333 SM1334 | SM1553 | SM0334 SM0333 | SM1353 |
| Colorless DNA ladders (dye not premixed) | SM0311 SM0312 | SM1331 SM1332 | SM1551 SM1552 | SM0331 SM0332 | SM1351 SM1352 |
| Base pairs (bp) | 250–10,000 | 75–20,000 | 100–5,000 | 100–10,000 | |
![]() | ![]() | ![]() | ![]() | ![]() |
* Bands represent each of the DNA fragments. The dark bands represent reference bands.
Related products
Explore other products for DNA electrophoresis workflow.
Molecular Biology Resource Library
Access webinars, videos, articles, and other resources on standard molecular biology techniques from the Thermo Scientific Library.
Nucleic Acid Electrophoresis Education
Learn about gel electrophoresis basics, workflow, considerations, applications, and troubleshooting in separation and analysis of nucleic acids.
Nucleic Acid Gel Electrophoresis and Blotting Support Center
Browse tips, troubleshooting help, and resources to optimize nucleic acid gel electrophoresis experiments.
Contact us
Email or call our technical application scientists for additional questions regarding DNA ladders.