Ladders can now be stored at room temperature for up to 24 months! During transition period you may receive packing indicating 6-month RT stability.
GeneRuler Ready-to-Use DNA Ladders
Thermo Scientific™

GeneRuler Ready-to-Use DNA Ladders

Thermo Scientific GeneRuler DNA Ladders offer a wide selection of size standard made with individual chromatography-purified fragments in the range of 10 bp to 48.5 kb.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeQuantitySize RangeNumber of Markers
SM03141KB DNA Ladder100 Applications250 to 10000 bp14
SM1213Ultra Low Range DNA Ladder100 Applications10 to 300 bp11
SM1193Low Range DNA Ladder100 Applications25 to 700 bp10
SM037350BP DNA Ladder100 Applications50 to 1000 bp13
SM13341KB Plus DNA Ladder100 Applications75 to 20000 bp15
SM0243100BP DNA Ladder100 Applications100 to 1000 bp10
SM0323100BP Plus DNA Ladder100 Applications100 to 3000 bp14
SM0334DNA Ladder Mix100 Applications100 to 1000 bp21
SM1353High Range DNA Ladder100 Applications10171 to 48502 bp8
SM13331KB Plus DNA Ladder500 Applications75 to 20000 bp15
SM0244100BP DNA Ladder500 Applications100 to 1000 bp10
SM0324100BP Plus DNA Ladder500 Applications100 to 3000 bp14
SM0333DNA Ladder Mix500 Applications100 to 1000 bp21
SM03131KB DNA Ladder500 Applications250 to 10000 bp14
Catalog number SM0314
Price (USD)
60.65
Online exclusive
73.00
Save 12.35
50 µg
Product Type:
1KB DNA Ladder
Quantity:
100 Applications
Size Range:
250 to 10000 bp
Number of Markers:
14
Request bulk or custom format
Price (USD)
60.65
Online exclusive
73.00
Save 12.35
50 µg

Thermo Scientific GeneRuler DNA Ladders serve as precise molecular weight markers for the sizing and approximate quantification of double-stranded DNA fragments, accommodating a range from 10 bp up to 48.5 kb. The uniform intensity of the bands, coupled with distinct bright reference bands, facilitates accurate sizing and orientation during gel electrophoresis. Furthermore, the defined quantity of DNA within each band enables the approximate quantification of experimental DNA samples.

GeneRuler DNA ladders are provided in a convenient ready-to-use format incorporating 6X TriTrack DNA Loading Dye (contains Xylene Cyanol FF, Bromophenol Blue, and Orange G). The three dye present in the buffer allow for easy tracking of DNA migration during electrophoresis.

Highlights

  • Wide selection of DNA ladders fitting a variety of experiments
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Easy visualization—three-colored loading dye for easy tracking of DNA migration during electrophoresis
  • Ready-to-use: premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months

Sustainability features

Ready-to-use GeneRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel CompatibilityAgarose Gels
Quantity100 Applications
Ready to LoadYes
Shelf Life24 months when stored at room temperature.
Volume (Metric)50 μg
Gel TypeAgarose
Label or DyeXylene Cyanol FF, Bromophenol Blue, Orange G
Number of Markers14
Product LineGeneRuler
Product Type1KB DNA Ladder
Size Range250 to 10000 bp
Unit Size50 µg

Frequently asked questions (FAQs)

Are the Thermo Scientific DNA ladders compatible with GelRed and GelGreen gel stains?

The GelRed or GelGreen nucleic acid dyes can be used with our DNA Ladders. Nucleic acid binding dyes can affect DNA migration during electrophoresis, therefore we highly recommend post-staining of gels. Please follow the gel staining protocol provided by the manufacturer of nucleic acid gel dyes.

Which Thermo Scientific DNA ladders are optimal for fast electrophoresis separation?

For fast sizing and quantification of double-stranded DNA, we recommend using FastRuler DNA Ladders. These ladders are easily resolved in a short separation distance after an 8-14 min run.

What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?

Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).

What are the end functional groups of Thermo Scientific DNA and RNA ladders?

Thermo Scientific DNA and RNA ladders have 5′-phosphate and 3′-hydroxyl groups.

Does the GeneRuler 1 kb DNA Ladder, ready-to-use have a fluorescent dye already mixed into it for detection on a gel?

No, this needs to be added to the gel.