AAV Purification Solutions for Gene Therapy Development
On this page:
Addressing purification challenges in AAV bioprocessing
Achieving high purity and yield during adeno-associated virus (AAV) downstream processing (DSP) requires overcoming several persistent challenges. Enriching full capsids from empty and partially filled capsids directly affects vector potency and dose consistency. Process-related impurities, such as host cell proteins (HCP), residual DNA, and adventitious viruses, as well as product-related impurities including empty, partially filled, and overfilled capsids, complicate workflows. Variability across AAV serotypes and engineered capsids demands flexible purification strategies. AAV purification solutions are designed to address these challenges while supporting productivity and scalability across development and cGMP manufacturing.
A workflow-based approach to AAV purification
AAV downstream processes typically integrate affinity-based capture followed by a non-affinity polishing step and analytics to support process consistency and manufacturing readiness.
Affinity chromatography selectively concentrates AAV particles while allowing many impurities to flow through. Subsequent polishing steps further refine purity following capture. Analytical methods are then used to characterize critical quality attributes to enhance process understanding and development decisions. 
Affinity capture strategies for AAV vectors
Affinity chromatography is the primary capture step following clarification in many AAV purification workflows.
Affinity ligands recognize specific epitopes on AAV capsid surfaces, allowing selective binding while unrelated impurities flow through. The ligand specificity can be serotype-specific by targeting proteins unique to its capsid, or it can target an epitope common to many AAV serotypes.
Affinity chromatography resins with broad serotype specificity can bind a broad range of natural and engineered AAV variants, although the latter may require confirmation of epitope preservation or further process optimization. High binding capacity at short residence times further supports processing efficiency.
Selectivity, throughput, and compatibility with downstream steps influence primary capture decisions when designing an AAV purification workflow. Affinity chromatography resins with high dynamic binding capacity (DBC) can process large volumes efficiently, reducing processing time. Load density, residence time, and buffer composition influence binding performance and recovery and should be optimized together.
Elution conditions need to release bound AAV particles while maintaining capsid integrity and minimizing aggregation, which may require serotype-specific excipients or neutralization strategies for aggregation-prone capsids. Compatibility with cleaning-in-place and resin reuse enables cost-effective manufacturing. Evaluating these factors during process development can help establish a scalable capture step aligned with downstream polishing requirements.
Polishing AAV vectors
Polishing AAV vectors for purity and consistency
Polishing addresses residual impurities and supports enrichment of full AAV capsids following capture, although no universal method exists. Separation conditions may require redevelopment for each serotype or construct. Anion exchange (AEX) chromatography can be used to reduce process- and product-related impurities, such as host cell proteins and empty capsids.
Chromatography conditions, including pH, conductivity, and excipients, can be tuned to influence impurity clearance and empty/full capsid separation. Optimizing these parameters during process development can help improve batch-to-batch consistency and overall AAV quality.
Learn more about AEX resins used for AAV polishing
High-throughput development tools
Speed AAV process development with high-throughput tools designed to rapidly screen and optimize purification conditions, from parallel resin condition screening in 96-well formats to flexible small-scale columns that streamline method development and scale-up. For early stage material generation and candidate screening, membrane-based affinity capture enables rapid AAV purification with significantly reduced processing times compared to traditional resin approaches.
96-well screening plates
A 96-well plate format can help streamline resin evaluation and screening by running multiple purifications in parallel. This approach enables the comparison of binding and wash conditions early, before proceeding to larger-scale purification studies.
Chromatography columns
Pre-packed columns simplify setup by reducing the need for on-site column packing. Formats range from bench-scale to process-scale and include resins for capture, intermediate, and polishing steps in purification workflows.
Membrane chromatography
Membrane chromatography offers higher flow rates and shorter processing times compared to traditional packed-bed resins and can help accelerate biologics production from development to manufacturing.
Scaling AAV purification toward cGMP manufacturing
Chromatography resins, chromatography column formats, and process robustness considerations support scale-up from development to cGMP manufacturing. Resin selection influences binding capacity, flow rate capability, and chemical stability across multiple cycles. Packed-bed columns offer flexibility for larger manufacturing volumes and established scale-up models, making them preferable over non-scalable approaches such as ultracentrifugation as programs advance toward cGMP manufacturing. Load density, residence time, and elution strategy should remain consistent across scales to maintain comparable purity and yield.
A portfolio supporting AAV purification workflows
AAV purification workflows benefit from integrated solutions across clarification, chromatography, and analytical characterization. Thermo Fisher offers affinity and anion exchange resins, prepacked columns, chromatography systems, and analytical assays.
These technologies allow for lysate clarification and particulate reduction prior to chromatography-based purification.
Anion exchange resins enhance polishing strategies and capsid enrichment within downstream workflows.
Analytical assays enable detection and quantification during purification and process development.
Integrating purification with downstream analytics
AAV purification process development works together with analytical method development. A robust purification process provides purified material for the development of qualified analytical methods, while analytics support process characterization and decision-making. Assay sensitivity and reproducibility are largely impacted by the purity of the standards used during analytical development. Likewise, reliable assays allow process scientists to quantify capsids and impurities while screening for and optimizing process conditions.
Analytical tools, including viral titer determination assays, ELISA kits, residual plasmid DNA quantification, and virus detection systems, support process characterization and impurity monitoring throughout development and manufacturing.
Not sure which resin is right for you?
The Bioprocess Resin Selection Guide is an interactive guide that helps scientists evaluate chromatography resins based on molecule type, application, and scale. Identify products that meet your purification goals and process needs by filtering data on resin chemistry, capacity, and operating conditions.
Frequently asked questions
Combining selective capture with appropriate polishing steps can help manage trade-offs between recovery and purity across the AAV purification process. Affinity chromatography enables high-purity capture while maintaining yield through selective binding and optimized elution conditions. Polishing steps using ion exchange chromatography (IEX) can then address residual impurities and enrich full capsids. Process parameters, including load density, gradient design, and pooling strategy, influence the balance between yield and purity . Establishing acceptable ranges for these parameters during development, guided by product quality requirements and analytical characterization, supports consistent performance across scales and manufacturing campaigns.
Affinity capture has increasingly become the primary capture step in many AAV workflows, reflecting a similar evolution to monoclonal antibody (mAb) purification, where Protein A (ProA) affinity chromatography became widely adopted for primary capture. Affinity is chosen for its high selectivity early in downstream processing, enabling significant impurity reduction and AAV recovery in a single step. Ion exchange chromatography plays a complementary role later in the workflow to address residual impurities and refine purity based on impurity profiles and process needs, particularly for full capsid enrichment and reduction of process-related contaminants.
Successful scale-up of AAV purification processes depends on resin selection, column scalability, and process robustness as workflows transition from development to cGMP manufacturing. Few chromatography formats scale with the same robustness or operational flexibility at larger volumes. Packed-bed resin-based approaches are often considered for long-term manufacturing readiness due to their scalability, mechanical stability, and compatibility with automated systems. Process parameters should remain consistent across scales, with loading conditions, residence times, and elution strategies verified using scale-down models. Emphasis on consistency, reproducibility, and compatibility with cGMP operations supports regulatory submissions and commercial manufacturing success.
While analytical results are essential for developing effective and robust downstream processes, high-purity materials are essential for developing sensitive, reproducible analytical methods, as impurities in reference standards can compromise assay accuracy and consistency. If impurities are not adequately removed, they may interfere with analytical readouts and increase variability in release results. Aligning purification and analytical strategies early in development can support more rapid process development, strengthen regulatory submissions, support well-defined specifications, and enable consistent long-term process control.
Chromatography and purification solutions that can support both current process development needs and longer-term manufacturing goals include affinity resins with broad serotype specificity, anion exchange resins optimized for full capsid enrichment, and chromatography column formats that enable scalable processing. Affinity chromatography using camelid antibody-based ligands can capture a wide range of AAV serotypes in a single platform. Anion exchange chromatography resins with large throughpores, like the POROS bead, are advantageous for AAV applications and enable full capsid enrichment and impurity clearance. Pre-packed chromatography columns support rapid process development and streamlined technology transfer, saving valuable time during manufacturing. These solutions can help address current purification challenges while establishing a foundation for scalable cGMP manufacturing.
Discover custom chromatography resin development
Custom chromatography resin development services can be an option for addressing AAV purification challenges when standard affinity or ion exchange solutions may not fully meet process needs.
Supporting other applications
Many chromatography-based purification principles used for AAV are also applied in related viral vectors and nucleic acid workflows. Affinity and ion exchange chromatography strategies can be adapted for other gene therapy vectors, including lentivirus (LV) and adenovirus applications.
Gene therapy bioprocessing solutions
Affinity chromatography resins
Chromatography and purification products 
POROS and CaptureSelect Chromatography
For research and development use only in support of FDA-regulated end uses. Not for diagnostic use or direct administration to humans or animals.