CaptureSelect™ HRP Anti-AAV9 Conjugate
Thermo Scientific™

CaptureSelect™ HRP Anti-AAV9 Conjugate

Thermo Scientific™ CaptureSelect™ HRP Anti-AAV9 Conjugate includes a 13-kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds adeno-associated virus particles of serotype 9 (AAV9).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7303332100100 μL
7303332500500 μL
Catalog number 7303332100
Price (USD)
4,235.00
Each
Quantity:
100 μL
Price (USD)
4,235.00
Each

CaptureSelect HRP Anti-AAV9 Conjugate includes a 13-kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds adeno-associated virus particles of serotype 9 (AAV9).

CaptureSelect HRP Anti-AAV9 Conjugate can be combined with CaptureSelect Biotin Anti-AAV9 Conjugate (Cat. No. 7103332100) in enzyme-linked immunoassays to quantify the total number of AAV9 capsids. Sandwich ELISA set-ups can be designed using the biotin anti-AAV9 conjugate as a capture reagent immobilized on a streptavidin coated surface. HRP anti-AAV9 conjugate is then used for detection of bound AAV9 capsids.

For assay development to measure total capsids for other AAV serotypes, we recommend CaptureSelect Biotin Anti-AAVX Conjugate (Cat. No. 7103522100) and CaptureSelect HRP Anti-AAVX Conjugate (Cat. No. 7303522100).

Product details

  • Format type: HRP, liquid
  • Buffer: PBS, 50% (v/v) glycerol
  • Storage: in freezer (-5 to -30°C)
  • Concentration: 0.5 mg/mL
  • Volume: 0.1 mL
  • Recommended working dilution for sandwich ELISA: 1:5000 to 1:10,000 (the optimal working dilution should be determined empirically for each application)

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications
Concentration0.5 mg/mL
For Use With (Application)Quantitation of AAV9 Capsids
FormulationLiquid, in PBS (7.4), 50% glycerol
Quantity100 μL
FormLiquid
Product LineCaptureSelect
TypeConjugate
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).