Search
Optimize your experiments to get the best results. We’ve compiled a detailed knowledgebase of the top tips and tricks to meet your research needs.
View the relevant questions below:
Beginning your experiment?
Visit our
No. After the addition of EDTA, there is an approximately 1:1 molar ratio of EDTA:Mg2+. EDTA chelates Mg2+ molecules on a 1:1 molar basis. Therefore, this RNA can be directly used in a reverse transcription reaction. First-strand reverse transcription buffers typically result in a final concentration of 2.5 mM Mg2+. If the reverse transcription buffer does not contain MgCl2, add it to the reaction to a final concentration of 2.25–2.5 mM.
You can run a no-RT control reaction with the SuperScript® VILO™ Master Mix by heat-inactivating the SuperScript® III Reverse Transcriptase enzyme for 5 min at 85°C. Then continue with the protocol as normal, using the mix that now has no RT activity.
It is not possible to directly measure the cDNA concentration by UV absorbance because the dNTPs from the reverse transcription step will also absorb and throw off the measurement. For an accurate reading, you would have to purify the cDNA first. Since this would lead to a loss of material, cDNA concentrations are estimated based on the initial RNA input instead. For example, if you used 100 ng of total RNA in a 20 µL reverse transcription reaction, you can assume a maximum of 100 ng of cDNA (at 5 ng/µL).
It may be possible to use your SYBR® Green primers for a TaqMan® assay, depending on how they were designed. You would have to design a separate probe to use with your existing primers. Please refer to the guidelines in this manual on “Manually Designing Primers and Probes” for the next steps. If you have Primer Express® Software, you can use that software to design a probe. Please note that restricting the design using the predesigned SYBR® primers may not allow for a successful probe design.
Since SYBR® Green can bind to any double-stranded DNA product, it is important to check your melt curves for the number of peaks. When primers are specific, you should see only one peak. Sometimes you may get extra peaks in a melt curve, which could be due to primer-dimers, a nonspecific product, or gDNA contamination. Please watch this short video for more information on multiple peaks in melt curves and how to deal with them.
Poor efficiency is typically caused by PCR inhibitors, limiting reagents, or suboptimal assay design. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool for more details. You can also review this short video.
There are several reasons that amplification could be delayed. Please see the information in our Real-Time Troubleshooting Tool for more details.
If your amplification curves do not have a normal appearance, you may be having an issue with the baseline settings. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool for more details.
There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool for more details.
There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool for more details.
In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them.
Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them.
If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:
In general, the target Tm of a TaqMan™ probe is 70°C. Using the Primer Express™ Software, you can check the Tm of a MGB or TAMRA™ probe probe.
Within the software, go to Primer Probe Test Tool and then copy/paste in your sequence of interest to see the Tm. If you need to check for a MGB based probe, make sure to choose Document Type: TaqMan™ MGB Quantification. If you need to check for a QSY® or TAMRA™ based probe, make sure to choose Document Type: TaqMan™ Quantification.
Yes, you can use Primer Express v3.0.1 on Win10 computers.
You can directly see all the primer and probe sequences from a test design right away using the Primer Express™ Software. Open the software and choose File => New. Choose either TaqMan™ MGB Quantification or TaqMan™ Quantification for Gene Expression Assay design. Copy/paste your gene sequence of interest into the new window and click on the green triangle to get primer/probe designs.
In Windows, please click on "Control Panel", then click on "Region and Language" tab. Under the "Administrative" tab, click on “Change System Locale” and ensure that the English language is selected. If this does not fix the problem, please email our technical support team at techsupport@lifetech.com for further assistance.
For Research Use Only. Not for use in diagnostic procedures.