Search
| Conjugate | Antibody | Host | Quantity | Cat. No. |
|---|---|---|---|---|
| Qdot® 525 | Anti-goat IgG | Donkey | 100 µL | Q22072 |
| Anti–mouse IgG | Donkey | 100 µL | Q22073 | |
| Goat | 200 µL | Q11041MP | ||
| Anti–rabbit IgG | Donkey | 100 µL | Q22074 | |
| Goat | 200 µL | Q11441MP | ||
| Qdot® 565 | Anti–goat IgG | Donkey | 100 µL | Q22075 |
| Anti–mouse IgG | Donkey | 100 µL | Q22076 | |
| Goat | 100 µL | Q11032MP | ||
| 200 µL | Q11031MP | |||
| Anti–rabbit IgG | Donkey | 100 µL | Q22077 | |
| Goat | 100 µL | Q11432MP | ||
| 200 µL | Q11431MP | |||
| Anti–rat IgG | Goat | 200 µL | Q11631MP | |
| Qdot® 585 | Anti–goat IgG | Donkey | 100 µL | Q22078 |
| Anti–mouse IgG | Donkey | 100 µL | Q22079 | |
| Goat | 200 µL | Q11011MP | ||
| Anti–rabbit IgG | Donkey | 100 µL | Q22080 | |
| Goat | 200 µL | Q11411MP | ||
| Qdot® 605 | Anti–goat IgG | Donkey | 100 µL | Q22081 |
| Anti–mouse IgG | Donkey | 100 µL | Q22082 | |
| Goat | 100 µL | Q11002MP | ||
| 200 µL | Q11001MP | |||
| Anti–rabbit IgG | Donkey | 100 µL | Q22083 | |
| Anti–rat IgG | Goat | 200 µL | Q11601MP | |
| Anti–human IgG | Goat | 200 µL | Q11201MP | |
| Qdot® 625 | Anti–goat IgG | Donkey | 100 µL | Q22084 |
| Anti–mouse IgG | Donkey | 100 µL | Q22085 | |
| Goat | 100 µL | A10195 | ||
| Anti–rabbit IgG | Donkey | 100 µL | Q22086 | |
| Goat | 100 µL | A10194 | ||
| Qdot® 655 | Anti–chicken IgY | Goat | 200 µL | Q14421MP |
| Anti–goat IgG | Donkey | 100 µL | Q22087 | |
| Rabbit | 200 µL | Q11821MP | ||
| Anti–mouse IgG | Donkey | 100 µL | Q22088 | |
| Goat | 100 µL | Q11022MP | ||
| 200 µL | Q11021MP | |||
| Anti–rabbit IgG | Donkey | 100 µL | Q22089 | |
| Goat | 100 µL | Q11422MP | ||
| 200 µL | Q11421MP | |||
| Anti–rat IgG | Goat | 200 µL | Q11621MP | |
| Anti–human IgG | Goat | 200 µL | Q11221MP | |
| Qdot® 705 | Anti–mouse IgG | Goat | 100 µL | Q11062MP |
| 200 µL | Q11061MP | |||
| Anti–rabbit IgG | Goat | 100 µL | Q11462MP | |
| 200 µL | Q11461MP | |||
| Qdot® 800 | Anti–mouse IgG | Goat | 200 µL | Q11071MP |
| Anti–rabbit IgG | Goat | 200 µL | Q11471MP | |
| All Qdot® nanocrystal secondary antibody conjugates are supplied as 1 µM solutions and are prepared either from whole IgG antibodies (in the case of the donkey antibody conjugates and the goat anti–chicken IgY conjugate) or from affinity-purified F(ab')2 fragments. | ||||
For Research Use Only. Not for use in diagnostic procedures.