Search
| BODIPY Dye | Abs * | Em * | COOH | STP | Succinimidyl Ester |
|---|---|---|---|---|---|
| BODIPY 493/503 | 500 | 506 | D2191 | ||
| BODIPY FL | 505 | 513 | D2183 (C3) D3834 (C5) | B10006 | D2184 (C3) D6140 (SSE) D6102 (X) D6184 (C5) |
| BODIPY R6G | 528 | 550 | D6180 | ||
| BODIPY 530/550 | 534 | 554 | D2187 | ||
| BODIPY TMR | 542 | 574 | D6117 (X) | ||
| BODIPY 558/568 | 558 | 569 | D2219 | ||
| BODIPY 564/570 | 565 | 571 | D2222 | ||
| BODIPY 576/589 | 576 | 590 | D2225 | ||
| BODIPY 581/591 | 584 | 592 | D2228 | ||
| BODIPY TR | 589 | 617 | D6116 (X) | ||
| BODIPY 630/650-X † | 625 | 640 | D10000 | ||
| BODIPY 650/655-X † | 646 | 660 | D10001 | ||
| * Approximate absorption (Abs) and fluorescence (Em) maxima, in nm, for the goat anti–mouse IgG antibody or dextran conjugates in aqueous buffer. † Not recommended for derivatizing proteins. (C3) = Propionic acid. (C5) = Pentanoic acid. (COOH) = Carboxylic acid.(SSE) = Sulfosuccinimidyl ester. (STP) = 4-Sulfotetrafluorophenyl ester. (X) = Aminohexanoyl spacer separating the dye and the SE. | |||||
For Research Use Only. Not for use in diagnostic procedures.