Table 14.2 Fluorescence characteristics of NeuroTrace fluorescent Nissl stains.
Cat. No. Fluorescent Color Ex * Em *
405 ± 20
445
≥450
485 ± 11
505
535 ± 17.5
510 ± 11.5
540
≥550
560 ± 20
590
610 ± 10
640 ± 10
660
682 ± 11
N21479Blue435455
+++
+
–
–
–
N21480Green500525
–
+++
+++
–
–
N21481Yellow515535
–
+++
+++
+++
–
N21482Red530615
–
–
++
+++
–
N21483Deep red640660
–
–
– ‡
–
++
* Approximate fluorescence excitation (Ex) and emission (Em) maxima, in nm. † Spectral characteristics in nm of the excitation filter, dichroic mirror and emission filter are shown top to bottom for each filter set. Relative signal strength for each filter combination is indicated with pluses and minuses. ‡ High background fluorescence.
For Research Use Only. Not for use in diagnostic procedures.