Table 18.1 Reactive oxygen species.
Reactive Oxygen SpeciesStructureDetection Reagents
Hydrogen peroxideH2O2
Hydroxyl radical *HO•
Hypochlorous acidHOCl
Nitric oxideNO
Peroxyl radical, including both alkylperoxyl and hydroperoxyl ref radicals (wherein R = H)ROO•
Peroxynitrite anion †ONOO–
Singlet oxygen ‡1O2
  • Singlet Oxygen Sensor Green reagent (S36002)
  • trans-1-(2'-methoxyvinyl)pyrene (M7913) ref
Superoxide anion•O2–
* Hydroxyl radicals can also be photosensitized by malachite green isothiocyanate (M689) or generated by a N-(1,10-phenanthrolin-5-yl)iodoacetamide (P6879) metal–ligand complex. † 3-Nitrotyrosine, a product of this potent nitrating reagent, can be detected with an anti-nitrotyrosine antibody (A21285). ‡ Singlet oxygen can also be photosensitized by hypericin (H7476), rose bengal diacetate (R14000) and merocyanine 540 (M24571).
For Research Use Only. Not for use in diagnostic procedures.