Technical Resources - Media Formulations
14287 - DPBS, calcium, magnesium, glucose, pyruvate
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0100.00.9009009
Magnesium Chloride (MgCl2-6H2O)203.0100.00.49261084
Potassium Chloride (KCl)75.0200.02.6666667
Potassium Phosphate monobasic (KH2PO4)136.0200.01.4705882
Sodium Chloride (NaCl)58.08000.0137.93103
Sodium Phosphate dibasic (Na2HPO4-7H2O)268.02160.08.059702
Other Components
D-Glucose (Dextrose)180.01000.05.5555553
Sodium Pyruvate110.036.00.3272727
Verweis
1. Dulbecco, R. and Vogt, M., (1954) Plaque formation and isolation of pure lines with Poliomyelitis viruses. J. Exp. Med., 98:167.
Related Products: