DPBS, calcium, magnesium, glucose, pyruvate
DPBS, calcium, magnesium, glucose, pyruvate
DPBS, calcium, magnesium, glucose, pyruvate
Gibco™

DPBS, calcium, magnesium, glucose, pyruvate

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
142870721000 mL
14287080500 mL
Catalog number 14287072
Price (USD)
65.65
Online exclusive
71.00
Save 5.35
Each
Quantity:
1000 mL
Customize this product
Price (USD)
65.65
Online exclusive
71.00
Save 5.35
Each

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.


This DPBS is modified as follows:
WithWithout
• Calcium• Phenol Red
• Magnesium 
• Glucose 
• Sodium Pyruvate 


The complete formulation is available.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialDPBS
IncludesSodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Recommended StorageStorage conditions: 2-8° C
Shelf life: 36 months from date of manufacture
SterilitySterile-filtered
Concentration1 X
For Use With (Application)Mammalian Cell Culture
Physical FormLiquid
Product LineGibco
Quantity1000 mL
Solution TypeDulbecco's Phosphate Buffered Saline
Unit SizeEach

Frequently asked questions (FAQs)

Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.