One-step reverse transcription-polymerase chain reaction (RT-PCR) is one of the most common techniques for RNA analysis. During RT-PCR, RNA molecules are first converted into complementary DNA (cDNA), which is then amplified by PCR. In the one-step RT-PCR approach, the reverse transcriptase and the DNA polymerase are premixed into a single tube, allowing both the RT and the subsequent PCR step to be performed in a single reaction. The advantages of one-step RT-PCR over two-step RT-PCR include fast and simple analysis, less pipetting steps, lower risk of errors and contamination, and suitability for high-throughput applications.
Learn about the differences between one-step and two-step RT-PCR
Invitrogen One-Step RT-PCR Systems Selection Guide
Table 1. Comparison of one-step RT-PCR systems.
Product photo. Three tubes and package.
SuperScript IV UniPrime One-Step RT-PCR System
Product photo. Three tubes and package.
SuperScript IV One-Step RT-PCR System
Product photo. Three tubes and package.
SuperScript III One-Step RT-PCR System with PlatinumTaqDNA Polymerase
Sensitivity0.01 pg0.01 pg0.01 pg
Range of amplicon length13 kb13.8 kb4.5 kb
Benchtop stability at RT24 h for up to 3 kb
4 h for ≥3 kb
Up to 4 h—
Resistance to inhibitors******
Multiplex******
RT time10 min10 min30 min
Extension time30 s/kb30 s/kb1 min/kb
Universal annealingYesNoNo
Tracking dyeYesNoNo
Integrated gDNA removalNoYes†No
 Order hereOrder hereOrder here
*Indicates degree of resistance to inhibitors and ability to multiplex
†A separate format with Invitrogen ezDNase Enzyme
Advantages of the SuperScript IV UniPrime One-Step RT-PCR System
  • Universal annealing temperature—does not require Tm optimization for each primer pair
  • Sensitivity and specificity—detect targets from low quantities of input RNA
  • Inhibitor resistance—achieve reliable results with low purity samples
  • Benchtop stability—allows room temperature reaction setup and preserves stability of assembled reactions
  • Amplicon length—synthesizes and amplifies a broad range of target lengths
  • Multiplex RT-PCR—co-cycle multiple targets without significant optimization
  • Colored reagents—allows visual tracking of reaction setup
The innovative universal annealing technology used in SuperScript IV UniPrime One-Step RT-PCR System enables primer annealing at 60°C for most primer pairs, helping minimize optimization steps and avoid mistakes in reaction setup (Figure 1).
Figure 1. One-step RT-PCR cycling under two annealing conditions. Nine targets were amplified from 10 ng Universal Human Reference RNA (UHRR); the first batch was amplified using a universal annealing temperature of 60°C (left), and the second batch was amplified using annealing temperatures calculated with the Tm calculator for Platinum SuperFi DNA Polymerase (right). The molecular weight marker is Thermo Scientific GeneRuler 1 kb Plus DNA Ladder, ready-to-use.
SuperScript IV UniPrime One-Step RT-PCR System is highly sensitive and detects low-abundance targets (Figure 2); this allows for one-step RT-PCR experiments with limited RNA input.
Gel image shows successful amplification of 0.43 kb fragment from serial dilution from 1 μg to 0.01 pg of UHRR with SuperScript IV UniPrime One-Step RT-PCR System.
 Click image to enlarge
Figure 2. Target detection from low amounts of input RNA. Amplification of 0.43 kb fragment from serial dilution from 1 μg to 0.01 pg of UHRR with SuperScript IV UniPrime One-step RT-PCR System. The molecular weight marker is Thermo Scientific GeneRuler 100 bp Plus DNA Ladder, ready-to-use.
SuperScript IV UniPrime One-Step RT-PCR System withstands the effect of common RT and PCR inhibitors such as copurified compounds from biological samples or reagents used in RNA purification. This exceptional robustness makes the system less dependent on RNA sample purity to help achieve reliable results. The enzymes in other one-step RT-PCR kits were inhibited by the presence of inhibitors (Figure 3).
Figure 3. Resistance to inhibitors. Detection of a 1 kb RNA target from 100 ng UHRR using the SuperScript IV One-step RT-PCR System or other one-step RT-PCR kits in reaction mixtures containing: (1) no inhibitor, (2) xylan (2.5 μg/μL), (3) humic acid (15 ng/μL), (4) SDS (0.013%), (5) Trozol (1.2%), or (6) LiCl (2.5 μg/μL).
With the innovative two-phase hot-start activation mechanism, the SuperScript IV UniPrime One-Step RT-PCR System allows for room temperature reaction setup and stability of preassembled reactions for an extended time. Highly efficient and specific amplification is observed even after prolonged storage at room temperature.
  • For targets up to 3 kb, successful amplification is observed after reactions are left for 24 h at room temperature (Figure 4A)
  • For longer targets, reactions can be stored for up to 4 hours (Figure 4B)
Figure 4A. Extended stability at room temperature. Using the SuperScript IV UniPrime One-Step RT-PCR System, one-step RT-PCR reactions were assembled with 1 kb RNA target from 1 ng–1 pg UHRR. One batch of assembled reactions was immediately loaded to the thermal cycler. The second batch of assembled reactions was kept at room temperature for 24 hours, and then loaded to the thermal cycler. Analysis of RT-PCR products shows that specific target amplification was achieved even after extended time at room temperature. NTC: no-template control. The molecular weight marker is Thermo Scientific GeneRuler 1 kb Plus DNA Ladder, ready-to-use.
Figure 4B. Extended stability at room temperature. Using the SuperScript IV UniPrime One-Step RT-PCR System, one-step RT-PCR reactions were assembled with 4.5 kb RNA target from 1 μg–1 pg of UHRR. One batch of assembled reactions was immediately loaded to the thermal cycler. The second batch of assembled reactions was kept at room temperature for 4 hours, and then loaded to the thermal cycler. Analysis of RT-PCR products shows that specific target amplification was achieved even after extended time at room temperature. NTC: no-template control. The molecular weight marker is Thermo Scientific GeneRuler 1 kb Plus DNA Ladder, ready-to-use.
Combination of highly processive enzymes in the SuperScript IV UniPrime One-Step RT-PCR System allows for the amplification of a broad range of target lengths (Figure 5). Full length cDNA is synthesized in as few as 10 min, and the PCR step requires an extension time of only 30 sec/kb, resulting in one of the shortest protocols among one-step RT-PCR kits.
Gel image showing detection of RNA fragments ranging between 0.2 and 9.4 kb. SuperScript IV UniPrime One-Step RT-PCR System provides high yield  and specificity across all target lengths.
 Click image to enlarge
Figure 5. Versatility across a broad range of target lengths. Detection of RNA fragments ranging from 0.2 to 9.4 kb with the SuperScript IV UniPrime One-Step RT-PCR System and other one-step RT-PCR kits.
With features such as high specificity, processivity, and universal annealing, the SuperScript IV UniPrime One-Step RT-PCR System can successfully multiplex without the need for significant optimization (Figure 6).
In colored format, the SuperScript IV RT Mix contains red tracking dye and the UniPrime RT-PCR Master mix contains blue tracking dye. When the two solutions are mixed during reaction setup, the final reaction mix turns purple, allowing for visual tracking of the reaction setup to prevent pipetting errors (Figure 7).
Wells containing UniPrime RT-PCR Master mix with blue tracking dye. Upon addition of SuperScript IV RT Mix containing red tracking dye, the reaction mix turns purple.
Figure 7. Reaction setup using colored format of the SuperScript IV UniPrime One-Step RT-PCR System.
Resources for SuperScript IV UniPrime One-Step RT-PCR System
Interactive selection tool: Find the right reverse transcriptase for your experiments.
Learn how the SuperScript IV UniPrime One-Step RT-PCR System makes it easy to set up reactions while bringing excellent results.
OEM one-step RT-PCR
To manufacture products according to your specifications, leverage our industry-leading expertise and infrastructure as a comprehensive third-party OEM service. We offer kitting, labeling, and commercialization services for one-step RT-PCR. As a vertically integrated organization, we control reagent quality throughout the nucleic acid synthesis process which reduces batch-to-batch variability.
To inquire about OEM one-step RT-PCR and other enzymes, contact us
Learn more:OEM & commercial supply services
One-Step RT-PCR Systems: Frequently asked questions
Q: Can I use SuperScipt IV One-Step RT-PCR System cycling protocol with the new SuperScript IV UniPrime One-Step RT-PCR System?
Yes. SuperScript IV UniPrime One-Step RT-PCR System works at both universal 60°C annealing temperature and at annealing temperature calculated with a Tm calculator.
Q: I am having difficulties performing one-step RT-PCR with long targets. What are your recommendations?
Good laboratory practices are important for long fragment one-step RT-PCR. These include using high-quality templates (pure, fresh, and intact) and fresh primer solutions. Optimization steps to consider include longer extension times as recommended in the protocols and increasing template amounts. Learn more about RT-PCR reaction optimization and setup by visiting our reverse transcription educational resources.
Q: Can the SuperScript IV UniPrime One-Step RT-PCR system be used with RNA targets with structural complexities?
With the SuperScript IV UniPrime One-Step RT-PCR system, cDNA synthesis can be performed at higher temperatures than with many other one-step RT-PCR products. For GC-rich or structurally complex RNA templates, it is recommended to increase the cDNA synthesis incubation temperatures up to 55–65°C.
Q: What is a two-phase hot-start activation mechanism?
The two-phase hot-start mechanism, utilized in SuperScript IV One-Step RT-PCR System and SuperScript IV UniPrime One-Step RT-PCR System, ensures sequential activation of RT and PCR enzymes in the one-step RT-PCR workflow. At ambient temperature, SuperScript IV RT remains inactive with a heat-sensitive RT-blocker. During the first hot-start activation phase at approximately 45°C, the RT-blocker is released, and the first-strand cDNA synthesis is initiated. During the second activation phase, the reaction is heated to 98°C to activate DNA Polymerase and simultaneously inactivate SuperScript IV RT. This mechanism separates the RT and PCR enzymes’ activities, delivering the highest RT-PCR specificity and yield.
Q: Can I perform TA cloning after amplifying an RNA target with the SuperScript IV UniPrime One-Step RT-PCR System?
The SuperScript IV UniPrime One-Step RT-PCR System produces blunt-end PCR products that can be cloned directly into blunt-end cloning vectors. TA cloning is also possible if 3′ dA-overhangs are added after PCR. Learn more about the use of different PCR enzymes for cloning application by visiting PCR educational resources.
Q: Will the tracking dyes in the colored format of SuperScript IV UniPrime One-Step RT-PCR System interfere with fragment analysis in E-Gel agarose gels?
Red and blue tracking dyes included in the colored format of SuperScript IV UniPrime One-Step RT-PCR System do not interfere with electrophoresis in E-Gel agarose gels. For optimal separation using E-Gel precast agarose gels, dilute the sample 2- to 30-fold, following the recommendations for specific E-gel.
Q: What DNA Polymerase is included in SuperScript IV UniPrime One-Step RT-PCR System?
SuperScript IV UniPrime One-Step RT-PCR System includes 2X UniPrime RT-PCR Master Mix containing modified Invitrogen Platinum SuperFi II DNA Polymerase, which provides high specificity, high yields and is ideally suited for applications that require sequence accuracy. Unlike other common proofreading enzymes, this polymerase is compatible with dUTP.
Q: Can I set up my reactions at room temperature using SuperScript IV UniPrime One-Step RT-PCR System?
Yes. Reaction setup with SuperScript IV UniPrime One-Step RT-PCR System can be performed at room temperature. Moreover, due to the innovative two-phase hot start mechanism, preassembled reactions remain stable at room temperature for up to 24 hours for targets up to 3 kb and up to 4 hours for targets >3 kb.
SuperScript One-Step RT-PCR Systems: Citations
The SuperScript One-Step RT-PCR Systems are frequently referenced in numerous peer-reviewed research publications and employed in various applications, including drug development, agriculture, infectious disease, and cancer research.
UseReference
One step RT-PCR was carried out with a tumor marker and control using miRNA extracted from tumor samples.Liu J, Zhang L, Wang Z et al. (2024) Prognostic value of miR-190a-5p in renal cell cancer and its regulatory effect on tumor progression. Int J Biol Markers 39(4):310–318. doi: 10.1177/03936155241290251. PMID: 39415706.
Synthesize cDNA and amplify DNA from human leukocyte samples prior to sequencing.Dominguez-Ortiz J, Álvarez-Gómez RM, Montiel-Manríquez R et al. (2024) A molecular characterization of the allelic expression of the BRCA1 founder Δ9-12 pathogenic variant and its potential clinical relevance in hereditary cancer. Int J Mol Sci 25(12):6773. doi: 10.3390/ijms25126773. PMID: 38928478.
Synthesize and amplify cDNA prior to qPCR.Saleh DO, Baraka SM, Jaleel GAA et al. (2024) Eugenol alleviates acrylamide-induced rat testicular toxicity by modulating AMPK/p-AKT/mTOR signaling pathway and blood-testis barrier remodeling. Sci Rep 14(1):1910. doi: 10.1038/s41598-024-52259-1. PMID: 38253778.
Analyze gene expression of nasopharyngeal carcinoma cells by reverse transcription-quantitative polymerase chain reaction.Wang Q, Yu Q, Liu Y (2023) E2F3 renders an immunosuppressive tumor microenvironment in nasopharyngeal carcinoma: involvements of the transcription activation of PRC1 and BIRC5. Immun Inflamm Dis 11(8):987. doi: 10.1002/iid3.987. PMID: 37647439.
RT-PCR of circRNA and mRNA from glioblastoma (GBM) cells.Zhang Y, Liu S, Wu C et al. (2024) Inhibition of circular JUN prevents the proliferation and invasion of glioblastoma via miR-3064-IGFBP5 axis. J Cell Mol Med 28(18):70098. doi: 10.1111/jcmm.70098. PMID: 39307884.
Used to clone the protein coding region of the ABCB1 mRNA (a transporter implicated in drug resistance).Hayashi A, Kamio K, Miyanaga A et al. (2024) Ivermectin enhances paclitaxel efficacy by overcoming resistance through modulation of ABCB1 in non-small cell lung cancer. Anticancer Res 44(12):5271–5282. doi: 10.21873/anticanres.17355. PMID: 39626921.
Synthesize cDNA and amplify DNA prior to qPCR for assessing levels of gene transcripts of DERL2.Liu L, Wu J, Yan Y et al. (2024) DERL2 (derlin 2) stabilizes BAG6 (BAG cochaperone 6) in chemotherapy resistance of cholangiocarcinoma. J Physiol Biochem 80(1):81–97. doi: 10.1007/s13105-023-00986-w. PMID: 37815698.
UseReference
cDNA was synthesized and amplified from plant RNA. Amplified fragments were cloned and sequenced.Angira A, Baranwal VK, Ranjan A et al. (2024) Optimization of DAC-ELISA and IC-RT-PCR using the developed polyclonal antibody and one-step RT-PCR assays for detection of Indian citrus ringspot virus in kinnow orange of Punjab, India. J Virol Methods 329:114972. doi: 10.1016/j.jviromet.2024.114972. PMID: 38880340.
cDNA synthesis and amplification from RNA isolated from plants. Amplified cDNA was used to create a library for sequencing on an Illumina system.Arndell T, Chen J, Sperschneider J et al. (2024) Pooled effector library screening in protoplasts rapidly identifies novel Avr genes. Nat Plants 10(4):572–580. doi: 10.1038/s41477-024-01641-y. PMID: 38409291.
Amplify a watermelon virus isolated from plant leaves. Amplified fragments were cloned and sequenced.Kwak HR, Hong SB, Kim JE et al. (2024) Construction and characterization of a full-length infectious cDNA clone of a strain of watermelon mosaic virus isolated from melon. Plant Pathol J 40(6):615–624. doi: 10.5423/PPJ.OA.07.2024.0103. PMID: 39639665.
To amplify the full genome of turtle grass virus X (TGVX). Also used in multiplex RT-PCR assays to confirm the absence of TGVX and any other potexviruses in the experimental plants.Alvarado-Marchena L, Furman BT, Breitbart M (2025) Construction and characterization of an infectious cDNA clone of turtle grass virus X from a naturally infected Thalassia testudinum plant. mBio 16(1):e0282824. doi: 10.1128/mbio.02828-24. PMID: 39660922.
UseReference
RT-PCR of Zika virus sample from a fetal biopsy sample prior to sequencing.Marquine S, Durand GA, Modenesi G et al. (2024) Sequence data from a travel-associated case of microcephaly highlight a persisting risk due to Zika virus circulation in Thailand. J Infect Dis 229(2):443–447. doi: 10.1093/infdis/jiad322. PMID: 37561039.
Nested PCR and sequencing from plasma samples.Manyana S, Pillay M, Gounder L et al. (2023) Affordable drug resistance genotyping of HIV-1 reverse transcriptase, protease and integrase genes, for resource limited settings. AIDS Res Ther 20(1):9. doi: 10.1186/s12981-023-00505-3. PMID: 36759801.
Create amplicons of the SARS-CoV-2 S gene to build a library for sequencing on an Illumina system.Darling TL, Harastani HH, Joshi A et al. (2024) Mucosal immunization with ChAd-SARS-CoV-2-S prevents sequential transmission of SARS-CoV-2 to unvaccinated hamsters. Sci Adv 10(31):eadp1290. doi: 10.1126/sciadv.adp1290. PMID: 39083604.
Used for library preparation for targeted amplicon sequencing.Caserta LC, Zhang J, Piñeyro P, Diel DG (2023) Rapid genotyping of porcine reproductive and respiratory syndrome virus (PRRSV) using MinION nanopore sequencing. PLoS One 18(5):e0282767. doi: 10.1371/journal.pone.0282767. PMID: 37220122.
Amplify a mutant strain of SARS-CoV-2 prior to sequencing.Zhang L, Xie X, Luo H et al. (2024) Resistance mechanisms of SARS-CoV-2 3CLpro to the non-covalent inhibitor WU-04. Cell Discov 10(1):40. doi: 10.1038/s41421-024-00673-0. PMID: 38594245.
Synthesize cDNA from 20-year-old cryopreserved samples.Dyer WB, Suzuki K, Levert A et al. (2024) Preservation of functionality, immunophenotype, and recovery of HIV RNA from PBMCs cryopreserved for more than 20 years. Front Immunol 15:1382711. doi: 10.3389/fimmu.2024.1382711. PMID: 39221258.
Influenza A whole-genome sequencing using Illumina and Nanopore systems.Goraichuk IV, Risalvato J, Pantin-Jackwood M et al. (2024) Improved influenza A whole-genome sequencing protocol. Front Cell Infect Microbiol 14:1497278. doi: 10.3389/fcimb.2024.1497278. PMID: 39669272.
Amplification of SARS-CoV-2 whole genome for Illumina sequencing.Botelho-Souza LF, Nogueira-Lima FS, Roca TP et al. (2021) SARS-CoV-2 genomic surveillance in Rondônia, Brazilian Western Amazon. Sci Rep 11(1):3770. doi: 10.1038/s41598-021-83203-2. PMID: 33580111.
UseReference
Used to map the 3’ UTR region of the mitochondrial and sperm isoform of TFAM.Lee W, Zamudio-Ochoa A, Buchel G et al. (2023) Molecular basis for maternal inheritance of human mitochondrial DNA. Nat Genet 55(10):1632–1639. doi: 10.1038/s41588-023-01505-9. PMID: 37723262.
Used for library preparation to screen short synthetic promoters in mammalian cells.Zahm AM, Owens WS, Himes SR et al. (2024) A massively parallel reporter assay library to screen short synthetic promoters in mammalian cells. Nat Commun 15(1):10353. doi: 10.1038/s41467-024-54502-9. PMID: 39609378.
UseReference
One step RT-PCR was carried out with a tumor marker and control using miRNA extracted from tumor samples.Liu J, Zhang L, Wang Z et al. (2024) Prognostic value of miR-190a-5p in renal cell cancer and its regulatory effect on tumor progression. Int J Biol Markers 39(4):310–318. doi: 10.1177/03936155241290251. PMID: 39415706.
Synthesize cDNA and amplify DNA from human leukocyte samples prior to sequencing.Dominguez-Ortiz J, Álvarez-Gómez RM, Montiel-Manríquez R et al. (2024) A molecular characterization of the allelic expression of the BRCA1 founder Δ9-12 pathogenic variant and its potential clinical relevance in hereditary cancer. Int J Mol Sci 25(12):6773. doi: 10.3390/ijms25126773. PMID: 38928478.
Synthesize and amplify cDNA prior to qPCR.Saleh DO, Baraka SM, Jaleel GAA et al. (2024) Eugenol alleviates acrylamide-induced rat testicular toxicity by modulating AMPK/p-AKT/mTOR signaling pathway and blood-testis barrier remodeling. Sci Rep 14(1):1910. doi: 10.1038/s41598-024-52259-1. PMID: 38253778.
Analyze gene expression of nasopharyngeal carcinoma cells by reverse transcription-quantitative polymerase chain reaction.Wang Q, Yu Q, Liu Y (2023) E2F3 renders an immunosuppressive tumor microenvironment in nasopharyngeal carcinoma: involvements of the transcription activation of PRC1 and BIRC5. Immun Inflamm Dis 11(8):987. doi: 10.1002/iid3.987. PMID: 37647439.
RT-PCR of circRNA and mRNA from glioblastoma (GBM) cells.Zhang Y, Liu S, Wu C et al. (2024) Inhibition of circular JUN prevents the proliferation and invasion of glioblastoma via miR-3064-IGFBP5 axis. J Cell Mol Med 28(18):70098. doi: 10.1111/jcmm.70098. PMID: 39307884.
Used to clone the protein coding region of the ABCB1 mRNA (a transporter implicated in drug resistance).Hayashi A, Kamio K, Miyanaga A et al. (2024) Ivermectin enhances paclitaxel efficacy by overcoming resistance through modulation of ABCB1 in non-small cell lung cancer. Anticancer Res 44(12):5271–5282. doi: 10.21873/anticanres.17355. PMID: 39626921.
Synthesize cDNA and amplify DNA prior to qPCR for assessing levels of gene transcripts of DERL2.Liu L, Wu J, Yan Y et al. (2024) DERL2 (derlin 2) stabilizes BAG6 (BAG cochaperone 6) in chemotherapy resistance of cholangiocarcinoma. J Physiol Biochem 80(1):81–97. doi: 10.1007/s13105-023-00986-w. PMID: 37815698.
UseReference
cDNA was synthesized and amplified from plant RNA. Amplified fragments were cloned and sequenced.Angira A, Baranwal VK, Ranjan A et al. (2024) Optimization of DAC-ELISA and IC-RT-PCR using the developed polyclonal antibody and one-step RT-PCR assays for detection of Indian citrus ringspot virus in kinnow orange of Punjab, India. J Virol Methods 329:114972. doi: 10.1016/j.jviromet.2024.114972. PMID: 38880340.
cDNA synthesis and amplification from RNA isolated from plants. Amplified cDNA was used to create a library for sequencing on an Illumina system.Arndell T, Chen J, Sperschneider J et al. (2024) Pooled effector library screening in protoplasts rapidly identifies novel Avr genes. Nat Plants 10(4):572–580. doi: 10.1038/s41477-024-01641-y. PMID: 38409291.
Amplify a watermelon virus isolated from plant leaves. Amplified fragments were cloned and sequenced.Kwak HR, Hong SB, Kim JE et al. (2024) Construction and characterization of a full-length infectious cDNA clone of a strain of watermelon mosaic virus isolated from melon. Plant Pathol J 40(6):615–624. doi: 10.5423/PPJ.OA.07.2024.0103. PMID: 39639665.
To amplify the full genome of turtle grass virus X (TGVX). Also used in multiplex RT-PCR assays to confirm the absence of TGVX and any other potexviruses in the experimental plants.Alvarado-Marchena L, Furman BT, Breitbart M (2025) Construction and characterization of an infectious cDNA clone of turtle grass virus X from a naturally infected Thalassia testudinum plant. mBio 16(1):e0282824. doi: 10.1128/mbio.02828-24. PMID: 39660922.
UseReference
RT-PCR of Zika virus sample from a fetal biopsy sample prior to sequencing.Marquine S, Durand GA, Modenesi G et al. (2024) Sequence data from a travel-associated case of microcephaly highlight a persisting risk due to Zika virus circulation in Thailand. J Infect Dis 229(2):443–447. doi: 10.1093/infdis/jiad322. PMID: 37561039.
Nested PCR and sequencing from plasma samples.Manyana S, Pillay M, Gounder L et al. (2023) Affordable drug resistance genotyping of HIV-1 reverse transcriptase, protease and integrase genes, for resource limited settings. AIDS Res Ther 20(1):9. doi: 10.1186/s12981-023-00505-3. PMID: 36759801.
Create amplicons of the SARS-CoV-2 S gene to build a library for sequencing on an Illumina system.Darling TL, Harastani HH, Joshi A et al. (2024) Mucosal immunization with ChAd-SARS-CoV-2-S prevents sequential transmission of SARS-CoV-2 to unvaccinated hamsters. Sci Adv 10(31):eadp1290. doi: 10.1126/sciadv.adp1290. PMID: 39083604.
Used for library preparation for targeted amplicon sequencing.Caserta LC, Zhang J, Piñeyro P, Diel DG (2023) Rapid genotyping of porcine reproductive and respiratory syndrome virus (PRRSV) using MinION nanopore sequencing. PLoS One 18(5):e0282767. doi: 10.1371/journal.pone.0282767. PMID: 37220122.
Amplify a mutant strain of SARS-CoV-2 prior to sequencing.Zhang L, Xie X, Luo H et al. (2024) Resistance mechanisms of SARS-CoV-2 3CLpro to the non-covalent inhibitor WU-04. Cell Discov 10(1):40. doi: 10.1038/s41421-024-00673-0. PMID: 38594245.
Synthesize cDNA from 20-year-old cryopreserved samples.Dyer WB, Suzuki K, Levert A et al. (2024) Preservation of functionality, immunophenotype, and recovery of HIV RNA from PBMCs cryopreserved for more than 20 years. Front Immunol 15:1382711. doi: 10.3389/fimmu.2024.1382711. PMID: 39221258.
Influenza A whole-genome sequencing using Illumina and Nanopore systems.Goraichuk IV, Risalvato J, Pantin-Jackwood M et al. (2024) Improved influenza A whole-genome sequencing protocol. Front Cell Infect Microbiol 14:1497278. doi: 10.3389/fcimb.2024.1497278. PMID: 39669272.
Amplification of SARS-CoV-2 whole genome for Illumina sequencing.Botelho-Souza LF, Nogueira-Lima FS, Roca TP et al. (2021) SARS-CoV-2 genomic surveillance in Rondônia, Brazilian Western Amazon. Sci Rep 11(1):3770. doi: 10.1038/s41598-021-83203-2. PMID: 33580111.
UseReference
Used to map the 3’ UTR region of the mitochondrial and sperm isoform of TFAM.Lee W, Zamudio-Ochoa A, Buchel G et al. (2023) Molecular basis for maternal inheritance of human mitochondrial DNA. Nat Genet 55(10):1632–1639. doi: 10.1038/s41588-023-01505-9. PMID: 37723262.
Used for library preparation to screen short synthetic promoters in mammalian cells.Zahm AM, Owens WS, Himes SR et al. (2024) A massively parallel reporter assay library to screen short synthetic promoters in mammalian cells. Nat Commun 15(1):10353. doi: 10.1038/s41467-024-54502-9. PMID: 39609378.
See how it performs in your lab
Request a free sample
Resources
Reverse transcriptase selection tool
Find the right products for your cDNA synthesis needs.
Reverse transcriptase webinars
Stay up-to-date on current industry trends in cDNA synthesis applications with on-demand webinars.
Reverse transcriptase videos
Watch instructional videos on troubleshooting tips, how-to videos, and explainers related to cDNA synthesis.
Reverse transcription education
cDNA Synthesis Protocol in Five Steps
Review a wealth of educational resources on cDNA synthesis.
Reverse transcriptase literature
Access white papers and technical and application notes for insights on cutting edge reverse transcription technology.
OEM and custom solutions
Contact us
PCR and cDNA Synthesis Support Center
Get support for your cDNA synthesis experiments.
Hero banner dark gradient overlay
Stylesheet for Classic Wide Template adjustments
© 2023 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.