Note, order lookup is only supported in the following countries: United States,
Canada.
If you are outside one of those countries please reach out to One Lambda Customer Support to submit an order status inquiry.
Use:Contains pre-calibrated ELISA reagents for the detection of masked HLA antibodies to 88 HLA Class I antigens in human serum. Each affinity-purified Class I single antigen is coated in one well of a Terasaki tray.
Storage Temperature:2° to 8° C
Package Includes:20 - trays (1 test/tray), Serum control (0.2 ml vial), Sterile deionized water (1 ml bottle), Alkaline phosphatase conjugated anti-human IgG 100X (0.3 ml), Antibody diluent (50 ml bottle), Wash buffer 10X (125 ml bottle), Colorimetric enzyme substrate (15 ml bottle), Stop reagent (25 ml bottle)
Please sign in to access your account. Customers must log in to access some content, recorded webinars, training materials, documents, and software.
Incorrect username or password
Need Help? Call website support Toll Free: +1 800 822 8824 option 2 or Email support team
Don’t have a profile?
Register Now
Registration
Please provide the following to setup your user account. If you do not know your center ID, please leave the center ID field blank.
This email may already be in use. Please reset your password or feel free to contact 1lambda-techsupport@thermofisher.com. Thank you.
Thank you for registering, please expect an email confirming your account registration status. For support please reach out to 1lambda-techsupport@thermofisher.com
Thank you for registering, please expect an email confirming your account registration status. For support please reach out to 1lambda-techsupport@thermofisher.com
Thank you for registering, please expect an email confirming your account registration status. For support please reach out to 1lambda-techsupport@thermofisher.com
Thank you for registering. Our team will verify your account and you will receive an email within 1-2 business days to confirm your registration status. For immediate support, please reach out to 1lambda-techsupport@thermofisher.com