SuperScript V VILO Master Mix with gDNA Removal, 50 rxns - FAQs
SuperScript V VILO Master Mix with gDNA Removal, 50 rxns - FAQs
View additional product information for SuperScript V VILO Master Mix - FAQs (11800004, 11800001, 11800111, 11800003, 11800113, 11800002, 11800112)
This product does not have any FAQs available.