AccuPrime Pfx SuperMix is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification.
Have Questions?
Catalog Number
Quantity
12344040
200 rxns
Catalog number 12344040
Price (USD)
809.65
Online exclusive
852.00
Save 42.35
Each
Quantity:
200 rxns
Request bulk or custom format
Price (USD)
809.65
Online exclusive
852.00
Save 42.35
Each
Invitrogen AccuPrime Pfx SuperMix provides reagents for high-fidelity, high-specificity amplification of DNA fragments. High fidelity is provided by a proprietary enzyme preparation containing recombinant DNA polymerase from Thermococcus species KOD with proofreading activity. This ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs is designed for efficient PCR amplification.
Features
>26-times higher fidelity than with Taq DNA polymerase
Reduced set-up time
Produces products with blunt ends
Applications
High-fidelity PCR
Amplification of fragments up to 15 kb
Cloning & mutagenesis
Notes
Thermostable AccuPrime Pfx SuperMix proteins enhance primer-template hybridization during every cycle, increasing specificity and yield.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)26X
Hot StartBuilt-In Hot Start
No. of Reactions200 Reactions
OverhangBlunt
PolymeraseAccuPrime Pfx DNA Polymerase
Quantity200 rxns
Reaction FormatSuperMix or Master Mix
Shipping ConditionWet or Dry Ice
Size (Final Product)15 kb or less
Concentration1.1X
Detection MethodPrimer-probe
For Use With (Application)Hot-start PCR, High-fidelity PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
AccuPrime Pfx SuperMix (4 × 1.125 mL)
Store at –20°C in a non-frost-free freezer.
Frequently asked questions (FAQs)
My oligonucleotide does not appear to be the right length when I checked by gel electrophoresis. Why is this?
Oligos should be run on a polyacrylamide gel containing 7 M urea and loaded with a 50% formamide solution to avoid compressions and secondary structures. Oligos of the same length and different compositions can electrophorese differently. dC's migrate fastest, followed by dA's, dT's, and then dG's. Oligos containing N's tend to run as a blurry band and generally have a problem with secondary structure.
The primers I am using worked for PCR initially, but over time, have stopped working. What happened?
Primers should be aliquoted for single use before PCR set-up. Heat just the aliquoted primers to 94 degrees for 1 min. Quick chill the primer on ice before adding to the PCR reaction. Some primers may anneal to themselves or curl up on themselves.
I don't see a pellet in my oligo tube order. Should I ask for a replacement?
The drying method dries the primer in a thin layer along the sidewalls of the tube instead of the bottom, therefore a pellet is not always visible and should still be ready to use.
There is a ball-shaped pellet at the bottom of my oligo tube. What is this and can I still use my oligo?
If the oligo was overheated, it will appear as a ball-shaped pellet attached to the bottom of the tube. This should not affect the quality of the oligo, and the oligo should be readily soluble in water.
There is a green color in my lyophilized oligo. Can I still use it?
If an oligo appears green in color, this is most likely due to ink falling into the tube. The oligo should still be fully functional. The color can be removed by doing an ethanol precipitation.
Quaternary epitope specificities of anti-HIV-1 neutralizing antibodies generated in rhesus macaques infected by the simian/human immunodeficiency virus SHIVSF162P4.
Authors:Robinson JE, Franco K, Elliott DH, Maher MJ, Reyna A, Montefiori DC, Zolla-Pazner S, Gorny MK, Kraft Z, Stamatatos L
Journal:J Virol
PubMed ID:20106929
'Monoclonal antibodies (MAbs) that neutralize human immunodeficiency virus type 1 (HIV-1) have been isolated from HIV-1-infected individuals or animals immunized with recombinant HIV-1 envelope (Env) glycoprotein constructs. The epitopes of these neutralizing antibodies (NAbs) were shown to be located on either the variable or conserved regions of the HIV-1 Env ... More
Ultraviolet-B-mediated induction of protein-protein interactions in mammalian cells.
Light-sensitive proteins are useful tools to control protein localization, activation and gene expression, but are currently limited to excitation with red or blue light. Here we report a novel optogenetic system based on the ultraviolet-B-dependent interaction of the Arabidopsis ultraviolet-B photoreceptor UVR8 with COP1 that can be performed in visible ... More