Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • On-demand videos
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • PDGFRA (CD140a) Antibodies

          Invitrogen

          CD140a (PDGFRA) Monoclonal Antibody (APA5), Biotin, eBioscience™

          Advanced Verification
          90 Published Figures
          121 References
          View all (76) PDGFRA (CD140a) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD140a (PDGFRA) Monoclonal Antibody (APA5), Biotin, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (90)
          • Advanced Verification (1)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 92

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD140a (PDGFRA) Antibody (13-1401-80) in Flow

          Staining of NIH/3T3 cells with 0.5 µg of Anti-Mouse CD140a (PDGF Receptor a) Biotin followed by Streptavidin PE (Product # 12-4317-87). Appropriate isotype controls were used (open histogram). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Western Blot (WB)
          CD140a (PDGFRA) Antibody in Western Blot, Flow Cytometry (WB, Flow)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          CD140a (PDGFRA) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          CD140a (PDGFRA) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunocytochemistry (ICC/IF)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD140a (PDGFRA) Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Immunohistochemistry (IHC)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody in Flow Cytometry (Flow)
          CD140a (PDGFRA) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          13-1401-80

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 21 publications 21 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 15 publications 15 publications

          Flow Cytometry (Flow)

          1 µg/test
          View 76 publications 76 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Affinity Purification (AP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Chicken, Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), Biotin, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          APA5

          Conjugate

          Biotin Biotin Biotin
          • Unconjugated
          • APC 
          • APC-eFluor 780
          • Brilliant UV 395
          • Brilliant UV 563
          • Brilliant UV 661
          • FITC
          • Functional Grade
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 702
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_466606

          Product Specific Information

          Description: The APA5 monoclonal antibody reacts with the mouse CD140a molecule, the alpha chain of the platelet derived growth factor receptor (PDGF receptor). PDGFRa is a receptor tyrosine kinase that forms dimers on the surface upon ligand binding and phosphorylates substrates. Dimers of PDGFR consist of either homodimers of alpha/alpha, beta/beta, or heterodimers of alpha/beta and serve as a substrate for its kinase activity. CD140a is expressed by embryonic tissues and mesenchymal-derived cells of the adult mouse tissues.

          Applications Reported: The APA5 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This APA5 antibody has been tested by flow cytometric analysis of NIH/3T3 cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Alpha platelet-derived growth factor receptor precursor (PDGF-R-alpha); alpha-type platelet-derived growth factor receptor; CD140 antigen-like family member A; MGC74795; OTTHUMP00000218656; PDGF A-chain; PDGF alpha chain; PDGF Receptor alpha; PDGF subunit A; PDGF-1; PDGF-R-alpha; pdgfr-a; PDGFR-alpha; unnamed protein product

          View more View less

          Gene Aliases: CD140a; Pdgfr-2

          View more View less

          UniProt ID: (Mouse) P26618

          View more View less

          Entrez Gene ID: (Mouse) 18595

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity platelet-derived growth factor alpha-receptor activity vascular endothelial growth factor-activated receptor activity platelet-derived growth factor receptor binding ATP binding vascular endothelial growth factor binding protein homodimerization activity phosphatidylinositol 3-kinase binding macromolecular complex binding platelet-derived growth factor binding phospholipase C activator activity
          Process(es)
          luteinization in utero embryonic development hematopoietic progenitor cell differentiation response to stress transmembrane receptor protein tyrosine kinase signaling pathway estrogen metabolic process positive regulation of cell proliferation male gonad development female gonad development anatomical structure morphogenesis organ morphogenesis negative regulation of platelet activation cell migration peptidyl-tyrosine phosphorylation signal transduction involved in regulation of gene expression extracellular matrix organization lung development adrenal gland development positive regulation of cell migration male genitalia development Leydig cell differentiation cellular response to reactive oxygen species platelet-derived growth factor receptor-alpha signaling pathway positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway wound healing odontogenesis of dentin-containing tooth protein autophosphorylation platelet-derived growth factor receptor signaling pathway positive regulation of fibroblast proliferation embryonic digestive tract morphogenesis embryonic cranial skeleton morphogenesis skeletal system morphogenesis positive regulation of peptidyl-tyrosine phosphorylation positive regulation of calcium-mediated signaling white fat cell differentiation positive regulation of chemotaxis cardiac myofibril assembly palate development face morphogenesis cell chemotaxis lung growth positive regulation of branching involved in lung morphogenesis retina vasculature development in camera-type eye positive regulation of ERK1 and ERK2 cascade platelet aggregation cellular response to amino acid stimulus metanephric glomerular capillary formation regulation of mesenchymal stem cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-vnqbn:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0