Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Description: The Connexin32 monoclonal antibody recognizes human, mouse, rat, and chicken Connexin 32. Connexins are a family of integral membrane proteins that form gap junction channels, which function to facilitate direct cell-cell communication. Gap junctions are comprised of two hemichannels, or connexons, which are made up of six connexin proteins. Over 20 connexin proteins have been identified and each combine to form either homomeric or heteromeric channels with different functional properties. Connexin 32 is expressed by myelin, neurons, oligodendrocytes, and hepatocytes, and plays a role in the development of ovarian follicles. Connexin 32 expression also plays a role in disease progression in chronic multiple sclerosis and experimental autoimmune encephalomyelitis. Mutations in the Connexin 32 gene, GJB1, result in a form of Charcot-Marie-Tooth disease which is characterized by peripheral nerve dysfunction.
Applications Reported: This Connexin32 antibody has been reported for use in western blotting, immunocytochemistry, immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.
Applications Tested: This Connexin32 antibody has been tested by immunohistochemistry of frozen mouse tissue and can be used at less than or equal to 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Filtration: 0.2 µm post-manufacturing filtered.
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-a gap junction protein; connexin 32; connexin g; connexin-32; connexin-43; GAP junction 28 kDa liver protein; gap junction membrane channel protein beta 1; gap junction protein, beta 1, 32kDa; unnamed protein product
Gene Aliases: CMTX; CMTX1; Cnx32; connexin-32; CX32; Cxng
UniProt ID: (Human) P08034, (Mouse) P28230, (Rat) P08033
Entrez Gene ID: (Human) 2705, (Chicken) 378797, (Mouse) 14618, (Rat) 29584
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support