Search
View additional product information for Nalgene™ Large Wide-Mouth Fluorinated HDPE Bottle with Closure - FAQs (2124-0005)
1 product FAQs found
All of our fluorinated products are processed to Level 5.