Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Proteins & Peptides ›
          • Thrombopoietin Proteins

          Gibco

          Human TPO (Thrombopoietin) Recombinant Protein, PeproTech®

          392 References
          View all (8) Thrombopoietin proteins

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Human TPO (Thrombopoietin) Recombinant Protein, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Testing Data (1)
          Human TPO (Thrombopoietin) Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.
          Human TPO (Thrombopoietin) Protein in Bioactivity (Bioactivity)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Human TPO (Thrombopoietin) Protein (300-18-100UG) in Bioactivity

          Bioassay analysis of Human TPO (Thrombopoietin) Recombinant Protein, PeproTech® (Product # 300-18-1MG). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Human TPO (Thrombopoietin) Protein in Bioactivity (Bioactivity)

          Product Details

          300-18-100UG

          Applications
          Tested Dilution
          Publications

          Neutralization (Neu)

          -
          View 1 publication 1 publication

          Control (Ctrl)

          Assay-dependent
          -

          In vitro Assay (In vitro)

          -
          View 353 publications 353 publications

          Miscellaneous PubMed (Misc)

          -
          View 34 publications 34 publications

          Western blot control (WB Ctrl)

          Assay-dependent
          -

          ELISA standard (ELISA Std)

          Assay-dependent
          -

          Bioactivity (Bioactivity)

          Assay-dependent
          -

          Cell culture (Cell Clt)

          -
          View 3 publications 3 publications

          Cell differentiation (Cell Diff)

          -
          - -

          Cell stimulation (Cell Stm)

          -
          - -

          Cell expansion (Cell Exp)

          -
          View 1 publication 1 publication
          Product Specifications

          Species

          Human

          Published species

          Hamster, Human, Monkey, Mouse, Non-human primate, Rat, Virus

          Expression System

          E. coli

          Amino acid sequence

          SPAPPACDLR VLSKLLRDSH VLHSRLSQCP EVHPLPTPVL LPAVDFSLGE WKTQMEETKA QDILGAVTLL LEGVMAARGQ LGPTCLSSLL GQLSGQVRLL LGALQSLLGT QLPPQGRTTA HKDPNAIFLS FQHLLRGKVR FLMLVGGSTL CVRRAPPTTA VPSRTSLVLT LNEL

          Molecular weight

          18.6 kDa

          Class

          Recombinant

          Type

          Protein

          Purity

          ≥ 98% by SDS-PAGE gel and HPLC analyses.

          Endotoxin concentration

          <1 EU/µg

          Activity

          The ED50 as determined by the dose-dependent stimulation of the proliferation of human MO7e cells was found to be ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          100 µg

          Purification

          purified

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          Product Specific Information

          Recombinant Human TPO is a fully biologically active 174 amino acid polypeptide (18.6 kDa), which contains the erythropoietin-like domain of the full length TPO protein.

          This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

          Target Information

          Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C-mpl ligand; Megakaryocyte colony-stimulating factor; Megakaryocyte growth and development factor; megakaryocyte stimulating factor; MGDF; ML; MPL ligand; Myeloproliferative leukemia virus oncogene ligand; prepro-thrombopoietin; Thrombopoietin; thrombopoietin nirs

          View more View less

          Gene Aliases: CAMT2; MGDF; MKCSF; ML; MPLLG; THC9; THCYT1; THPO; TPO

          View more View less

          UniProt ID: (Human) B7ZLR8

          View more View less

          Entrez Gene ID: (Human) 7066

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline