Pierce™ PNPP Substrate Kit, 105 tablets kit - FAQs
Pierce™ PNPP Substrate Kit, 105 tablets kit - FAQs
View additional product information for PNPP Substrates - FAQs (34047, 34045, 37620, 37621)
1 product FAQs found
What alkaline phosphatase substrates do you offer?