Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • On-demand videos
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • IL-12 p70 Antibodies

          Invitrogen

          IL-12 p70 Polyclonal Antibody, PeproTech®

          View all (21) IL-12 p70 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite IL-12 p70 Polyclonal Antibody, PeproTech®

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          IL-12 p70 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IL-12 p70 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          IL-12 p70 Antibody (500-P154HG-100UG) in WB

          Western Blot: To detect Human IL-12 by Western Blot analysis IL-12 Polyclonal Antibody (Product # 500-P154HG-1MG) can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-12 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          IL-12 p70 Antibody in Western Blot (WB)
          IL-12 p70 Antibody in Western Blot (WB)
          IL-12 p70 Antibody in ELISA (ELISA)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          IL-12 p70 Polyclonal Antibody, PeproTech®

          Product Details

          500-P154HG-100UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          ELISA (ELISA)

          0.5-2.0 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Goat

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          HEK293 cells-derived Recombinant Human IL-12 p70
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2942209

          Product Specific Information

          AA Sequence of recombinant protein: p35 Subunit: RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNAS. p40 Subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS

          Preparation: Produced from sera of goats immunized with highly pure Recombinant Human IL-12. Anti-Human IL-12-specific antibody was purified by affinity chromatography employing an immobilized Human IL-12 matrix.

          Sandwich ELISA: To detect Human IL-12 by sandwich ELISA (using 100 µL/well) a concentration of 0.5-2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IL-12 (500-P154HGBT) as a detection antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human IL-12.

          Western Blot: To detect Human IL-12 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-12 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          Target Information

          IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 20C2; CLMF p35; CLMF p40; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12p70; IL-12 p75; IL-12 subunit p40; IL-12, subunit p35; IL12; IL12, subunit p40; IL12A/B; IL12AB; IL35 subunit; ILN; Interleukin; interleukin 12, p35; interleukin 12, p40; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin-12 alpha chain; interleukin-12 beta chain; Interleukin-12 p70; MGC151228; MGC151232; natural killer cell stimulatory factor 2; cytotoxic lymphocyte maturation factor 2; IL12p40; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF/CLMF; IL12p35; 35kD; NKSF/CLMF; IL12p40; 40kD

          View more View less

          Gene Aliases: CLMF; CLMF2; IL-12A; IL-12B; IMD28; IMD29; NFSK; NKSF; NKSF1; NKSF2; P35

          View more View less

          UniProt ID: (Human) P29459, (Human) P29460

          View more View less

          Entrez Gene ID: (Human) 3592, (Human) 3593

          View more View less

          Function(s)
          cytokine activity interleukin-12 receptor binding protein binding growth factor activity interleukin-12 beta subunit binding interleukin-27 binding protein heterodimerization activity cytokine receptor activity cytokine receptor binding interleukin-12 alpha subunit binding identical protein binding macromolecular complex binding interleukin-23 receptor binding
          Process(es)
          regulation of cytokine production positive regulation of leukocyte mediated cytotoxicity positive regulation of T cell mediated cytotoxicity positive regulation of defense response to virus by host natural killer cell activation involved in immune response positive regulation of lymphocyte mediated immunity positive regulation of T-helper 1 type immune response positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target immune response JAK-STAT cascade response to UV-B cell migration natural killer cell activation negative regulation of interleukin-10 production negative regulation of interleukin-17 production positive regulation of granulocyte macrophage colony-stimulating factor production positive regulation of interferon-gamma production positive regulation of interleukin-10 production positive regulation of interleukin-12 production positive regulation of interleukin-17 production positive regulation of tumor necrosis factor production positive regulation of natural killer cell activation positive regulation of natural killer cell proliferation positive regulation of mononuclear cell proliferation positive regulation of tissue remodeling positive regulation of smooth muscle cell apoptotic process interleukin-12-mediated signaling pathway interleukin-23-mediated signaling pathway T-helper 1 type immune response T-helper cell differentiation positive regulation of T cell proliferation positive regulation of activated T cell proliferation positive regulation of memory T cell differentiation positive regulation of osteoclast differentiation positive regulation of cell adhesion positive regulation of JAK-STAT cascade oocyte development negative regulation of smooth muscle cell proliferation positive regulation of lymphocyte proliferation negative regulation of protein secretion regulation of inflammatory response positive regulation of inflammatory response defense response to Gram-negative bacterium positive regulation of NK T cell activation positive regulation of NK T cell proliferation negative regulation of vascular endothelial growth factor signaling pathway positive regulation of NIK/NF-kappaB signaling negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis positive regulation of T-helper 17 type immune response positive regulation of T-helper 17 cell lineage commitment negative regulation of T-helper 1 type immune response response to virus regulation of B cell proliferation response to lipopolysaccharide positive regulation of tyrosine phosphorylation of STAT protein positive regulation of natural killer cell mediated cytotoxicity defense response to Gram-positive bacterium interleukin-35-mediated signaling pathway extrinsic apoptotic signaling pathway cellular response to virus positive regulation of dendritic cell chemotaxis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-74d7b4b4d9-ncdd4:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline