Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Podoplanin Antibodies

          Invitrogen

          Podoplanin Monoclonal Antibody (eBio8.1.1 (8.1.1)), Alexa Fluor™ 488, eBioscience™

          Advanced Verification
          32 Published Figures
          41 References
          View all (76) Podoplanin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Podoplanin Monoclonal Antibody (eBio8.1.1 (8.1.1)), Alexa Fluor™ 488, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (32)
          • Advanced Verification (5)
          Podoplanin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Podoplanin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 43

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Podoplanin Antibody (53-5381-80) in IHC (P)

          Immunohistochemical analysis of Podoplanin was performed using formalin-fixed paraffin-embedded mouse lung and mouse skeletal muscle tissue sections. To expose the target protein, heat-induced epitope retrieval was performed on de-paraffinized sections using eBioscience™ IHC Antigen Retrieval Solution - High pH (10X) (Product # 00-4956-58) diluted to 1X solution in water in a decloaking chamber at 110 degre... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Podoplanin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Podoplanin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Podoplanin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Podoplanin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Podoplanin Antibody in Western Blot (WB)
          Podoplanin Antibody in Flow Cytometry (Flow)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (PFA fixed), Immunocytochemistry, Flow Cytometry (IHC (PFA), ICC/IF, Flow)
          Podoplanin Antibody in Immunocytochemistry (ICC/IF)
          Podoplanin Antibody in Immunocytochemistry (ICC/IF)
          Podoplanin Antibody in Immunocytochemistry (ICC/IF)
          Podoplanin Antibody in Immunocytochemistry (ICC/IF)
          Podoplanin Antibody in Immunocytochemistry (ICC/IF)
          Podoplanin Antibody in Immunocytochemistry (ICC/IF)
          Podoplanin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Podoplanin Antibody in Immunocytochemistry (ICC/IF)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Immunohistochemistry (IHC)
          Podoplanin Antibody in Flow Cytometry (Flow)
          Podoplanin Antibody in Flow Cytometry (Flow)
          Podoplanin Antibody in Flow Cytometry (Flow)
          Podoplanin Antibody in Flow Cytometry (Flow)
          Podoplanin Antibody in Flow Cytometry (Flow)
          Podoplanin Antibody in Flow Cytometry (Flow)
          Podoplanin Antibody in Flow Cytometry (Flow)
          Podoplanin Antibody in Flow Cytometry (Flow)
          Podoplanin Antibody in Flow Cytometry (Flow)
          Podoplanin Antibody
          Podoplanin Antibody
          Podoplanin Antibody
          Podoplanin Antibody
          Podoplanin Antibody

          Product Details

          53-5381-80

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          -

          Immunohistochemistry (IHC)

          -
          View 11 publications 11 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          5 µg/mL
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 8 publications 8 publications

          Flow Cytometry (Flow)

          0.5 µg/test
          View 21 publications 21 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Hamster, Human, Mouse, Rat

          Host/Isotype

          Syrian hamster / IgG

          Recommended Isotype Control

          Syrian Hamster IgG Isotype Control, Alexa Fluor™ 488, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio8.1.1 (8.1.1)

          Conjugate

          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • Unconjugated
          • Biotin 
          • eFluor 660
          • PE 
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 600
          • Request custom conjugation

          Excitation/Emission Max

          499/520 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1106991

          Product Specific Information

          Description: The 8.1.1 monoclonal antibody reacts with mouse podoplanin (T1a, gp38, aggrus), a 43 kDa transmembrane glycoprotein, named for its expression in kidney glomerular epithelial cells (podocytes). In addition, Podoplanin is expressed in epithelial and mesothelial cells such as intestinal epithelium, alveolar type I cells, podocytes, and mesothelium of the visceral peritoneum. It was also shown to be a potent marker for lymphatic endothelium. Podoplanin is also expressed by subcapsular epithelial cells of the murine thymus. Mice deficient in Podoplanin die at birth because of a respiratory defect and congenital lymphedema due to a failure in lymphatic pattern formation.

          Applications Reported: This eBio8.1.1 (8.1.1) antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This eBio8.1.1 (8.1.1) antibody has been tested by flow cytometric analysis of the TE-71 cell line. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Podoplanin (PDPN, T1A, gp38, Aggrus) is a mucin type-1 glycoprotein with a 40-43 kDa molecular weight. Podoplanin is expressed in many tumors and normal cells, especially lymphatic epithelial cells and follicular DCs. Podoplanin localizes in stromal cells of peripheral lymphoid tissue and thymic epithelial cells. As a regulator of the lymphatic endothelium, podoplanin probably plays a role in maintaining the unique shape of podocytes. Podoplanin appears to serve as a ligand for CLEC-2 and expression is positively correlated with tumors expressing greater invasive and metastatic potential. Podoplanin is directly involved in cell migration, aids metastases formation and tumor cell invasion of tissue. Further, it has also been determined that podoplanin expression is induced through Src activation of Cas and, recently, there are indications of podoplanin presence on Th17 cells. Initial studies below indicate the presence of podoplanin on a small but distinct population of CD4+ cells from human PBMC. Dysfunction of the podoplanin protein has been linked to diseases such as lymphangioma and subglottis neoplasm. Podoplanin has also been proposed as a marker for lung injury due to the physiological function of the protein.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: aggrus; CTA-520D8.1; E11; Glycoprotein 38; Gp38; OTS-8; OTTHUMP00000009640; OTTHUMP00000044504; PA2.26 antigen; Podoplanin; RP23-348F1.2; Transmembrane glycoprotein E11

          View more View less

          Gene Aliases: Gp38; OTS-8; Ots8; Pdpn; RANDAM-2; T1-alpha; T1a; T1alpha

          View more View less

          UniProt ID: (Mouse) Q62011

          View more View less

          Entrez Gene ID: (Mouse) 14726

          View more View less

          Function(s)
          water transmembrane transporter activity folic acid transporter activity amino acid transmembrane transporter activity water channel activity
          Process(es)
          cell morphogenesis lymphangiogenesis prostaglandin metabolic process water transport amino acid transport movement of cell or subcellular component cell adhesion signal transduction multicellular organism development cell proliferation regulation of cell shape positive regulation of platelet activation folic acid transport single organismal cell-cell adhesion lung development positive regulation of cell migration tube morphogenesis lung alveolus development positive regulation of cellular component movement visceral serous pericardium development regulation of G1/S transition of mitotic cell cycle
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline