Product Image
Thermo Scientific™

ABsolute QPCR Mix, ROX

**********************Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’veRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
AB1138B1600 Reactions
AB1139A
also known as AB-1139/A
400 Reactions
Catalog number AB1138B
Price (USD)
3,490.00
Each
No. of Reactions:
1600 Reactions
Price (USD)
3,490.00
Each
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of ABsolute QPCR Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute qPCR Mixes are optimized for use with hybridization probes and contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes. The 2X master mix contains a proprietary reaction buffer that provides high sensitivity, specificity, and reproducibility. ABsolute qPCR Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, and with no ROX reference dye. There is also a master mix version optimized for capillary instruments.

Highlights

• High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
• Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency
• Optimised buffer - For maximal performance in multiplex reactions

Applications
• Gene expression analysis
• siRNA validation
• Genotyping

Related Products
ABsolute QPCR Mix, no ROX
ABsolute QPCR Mix, low ROX
ABsolute QPCR Mix, ROX
ABsolute QPCR Capillary Mix
ABsolute QPCR Mix, SYBR Green, no ROX
ABsolute QPCR Mix, SYBR Green, low ROX
ABsolute QPCR Mix, SYBR Green, ROX
ABsolute QPCR Mix, SYBR Green, fluorescein
ABsolute QPCR Capillary Mix, SYBR Green
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)2 X
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
No. of Reactions1600 Reactions
PolymeraseTaq DNA Polymerase
Product LineABsolute
Product TypeqPCR Mix
Quantity1,600 reactions
Sample TypeDNA, dsDNA
Sufficient For1600 Reactions
Volume20 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains:
• 16 x 1.25 mL ABsolute QPCR ROX Mix (2X)

Store at -20°C