Product Image
Thermo Scientific™

ABsolute QPCR Mix, SYBR Green, no ROX

**********************Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호반응 수
AB1159A
AB-1159/A으로도 사용됨
400 Reactions
AB1158B
AB-1158/B으로도 사용됨
1,600 Reactions
카탈로그 번호 AB1159A
AB-1159/A으로도 사용됨
제품 가격(KRW)
757,000
온라인 행사
Ends: 30-Jun-2026
841,000
할인액 84,000 (10%)
Each
반응 수:
400 Reactions
제품 가격(KRW)
757,000
온라인 행사
Ends: 30-Jun-2026
841,000
할인액 84,000 (10%)
Each
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of ABsolute QPCR Mix, SYBR Green, and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute qPCR SYBR Green Mixes are optimized for SYBR Green chemistry and contain all the components necessary to perform quantitative PCR, with the exception of template and primers. The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets.

ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, Fluorescein or no reference dye. A master mix optimized for capillary instruments is also available.

Highlights

• High Sensitivity & Specificity—The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
• Optimised buffer containing SYBR Green I
• Reproducibility—All master mixes undergo extensive QC to ensure batch to batch consistency
• Broad dynamic range

Applications

• Gene expression analysis
• siRNA validation
• Genotyping

Related Products
ABsolute QPCR Mix, no ROX
ABsolute QPCR Mix, low ROX
ABsolute QPCR Mix, ROX
ABsolute QPCR Capillary Mix
ABsolute QPCR Mix, SYBR Green, no ROX
ABsolute QPCR Mix, SYBR Green, low ROX
ABsolute QPCR Mix, SYBR Green, ROX
ABsolute QPCR Mix, SYBR Green, fluorescein
ABsolute QPCR Capillary Mix, SYBR Green
For Research Use Only. Not for use in diagnostic procedures.
사양
호환성qPCR Cyclers
용도(장비)BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene
반응 수400 Reactions
중합효소Taq DNA Polymerase
제품라인ABsolute
제품 유형Real Time PCR SYBR Master Mix
수량400 reactions
샘플 종류DNA, dsDNA
충분400 Reactions of 25 μL Each
부피5 mL
농도2X
검출 방법SYBR
용도(애플리케이션)Gene Expression
GC-Rich PCR PerformanceHigh
PCR 방법qPCR
반응 속도Standard
Unit SizeEach
구성 및 보관
Contains:
• 1 x 5 mL ABsolute QPCR SYBR Green Mix (2X)
• 100 μL MgCl2 (1M)
Sufficient for 400 x 25 μL reactions.

Store at -20°C