Product Image
Thermo Scientific™

ABsolute QPCR Mix, ROX

**********************Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve詳細を見る
製品番号(カタログ番号)反応数
AB1139A
または、製品番号AB-1139/A
反応400回分
AB1138B1600 Reactions
製品番号(カタログ番号) AB1139A
または、製品番号AB-1139/A
価格(JPY)
77,900
Each
反応数:
反応400回分
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of ABsolute QPCR Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute qPCR Mixes are optimized for use with hybridization probes and contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes. The 2X master mix contains a proprietary reaction buffer that provides high sensitivity, specificity, and reproducibility. ABsolute qPCR Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, and with no ROX reference dye. There is also a master mix version optimized for capillary instruments.

Highlights

• High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
• Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency
• Optimised buffer - For maximal performance in multiplex reactions

Applications
• Gene expression analysis
• siRNA validation
• Genotyping

Related Products
ABsolute QPCR Mix, no ROX
ABsolute QPCR Mix, low ROX
ABsolute QPCR Mix, ROX
ABsolute QPCR Capillary Mix
ABsolute QPCR Mix, SYBR Green, no ROX
ABsolute QPCR Mix, SYBR Green, low ROX
ABsolute QPCR Mix, SYBR Green, ROX
ABsolute QPCR Mix, SYBR Green, fluorescein
ABsolute QPCR Capillary Mix, SYBR Green
研究用途にのみご使用ください。診断目的には使用できません。
仕様
フィデリティ(vs. Taq)2 X
使用対象 (装置)7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
反応数反応400回分
ポリメラーゼTaq DNA Polymerase
製品ラインABsolute
製品タイプqPCR Mix
数量400 reactions
サンプルタイプDNA, dsDNA
対応可能対象400反応
容量5 mL
濃度2X
検出法Primer-probe
使用対象(アプリケーション)遺伝子発現解析
GC-Rich PCR Performance高
PCR法qPCR
反応速度スタンダード
Unit SizeEach
組成および保存条件
Contains:
• 1 x 5 mL ABsolute QPCR ROX Mix (2X)

Store at -20°C