EnzChek™ Reverse Transcriptase Assay Kit - FAQs
EnzChek™ Reverse Transcriptase Assay Kit - FAQs
View additional product information for EnzChek™ Reverse Transcriptase Assay Kit - FAQs (E22064)
2 product FAQs found
With the EnzChek Reverse Transcriptase Assay Kit, how do I expand the dynamic range?
To expand the upper end of the PicoGreen’s dynamic range, simply increase dye concentration. By doubling the concentration, the dynamic range will double, etc. Keep in mind, however that increasing the dye concentration sacrifices the lower end of the assay due to increasing background fluorescence.
Does the assay EnzChek Reverse Transcriptase Assay Kit (Cat. No. E22064) work on retroviruses other than HIV?
The assay should work with any reverse transcriptase.
Note: This is a solution reaction, so the reverse transcriptase needs to be available in solution to work, not encapsulated.