Phusion™ Plus DNA Polymerase, 100 x 50 μL reactions - FAQs
Phusion™ Plus DNA Polymerase, 100 x 50 μL reactions - FAQs
View additional product information for Phusion™ Plus DNA Polymerase - FAQs (F630S, F630XL, F630L)
2 product FAQs found
What is the fidelity of the Phusion Plus DNA Polymerase (Cat. No. F630S)?
Please see "Conclusions" within the technical note for the Phusion Plus DNA Polymerase (Cat. No. F630S) (see link: https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/measuring-error-rates-ngs-dna-polymerases-tech-note.pdf).
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.
Can I set up my reactions on ice if I am using the Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix (Cat. No. F631S)?
Reaction setup with Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix can be done either on ice or at room temperature.