Phusion™ Plus DNA Polymerase
Thermo Scientific™

Phusion™ Plus DNA Polymerase

Thermo Scientific Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
F630S100 Reactions
F630L500 Reactions
F630XL2000 Reactions
Catalog number F630S
Price (USD)
139.65
Online exclusive
160.00
Save 20.35
Each
No. of Reactions:
100 Reactions
Request bulk or custom format
Price (USD)
139.65
Online exclusive
160.00
Save 20.35
Each
Thermo Scientific Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing. It is a Pyrococcus-like proofreading DNA polymerase fused to a processivity-enhancing domain, enabling Phusion Plus DNA Polymerase to generate PCR sequences with high accuracy, sensitivity, and inhibitor tolerance.

Features include:

  • >100X higher sequence accuracy than Taq polymerase
  • Universal annealing temperature of 60°C in PCR
  • Hot-start mechanism reduces non-specific amplification and primer degradation
  • Automation enabled due to 24-hour stability of assembled reactions
  • High inhibitor tolerance for work with DNA of suboptimal purity

The novel Phusion Plus reaction buffer includes proprietary additives that stabilize primer/template duplexes during the annealing step at 60°C. This unique buffer formulation increases PCR specificity without the need to optimize annealing temperature for each primer pair, helping save time and avoid mistakes in PCR runs. Furthermore, with Phusion Plus DNA Polymerase and its universal annealing feature, different PCR reactions can be cycled together using the same protocol.

Phusion Plus DNA Polymerase is inactive at ambient temperatures due to its hot-start mechanism mediated by Affibody molecules, allowing setup and storage of assembled PCR reactions at room temperature. Enzyme activity is fully restored after the initial denaturation step.

Applications:

  • High-fidelity PCR
  • Molecular cloning and subcloning
  • Template generation for sequencing
  • GC-rich PCR
  • Long-range PCR
  • Multiplex target amplification
  • Synthetic biology and DNA assembly workflows
  • DNA template generation for in vitro transcription and mRNA synthesis
  • Site-directed mutagenesis
  • Microbial genomics and microbiome research
  • Synthetic biology and DNA assembly workflows

Notes

  • To simplify reactrion setup, Phusion Plus DNA Polymerase is also available in ready-to-use master mix formats.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)>100X
Hot StartBuilt-In Hot Start
No. of Reactions100 Reactions
OverhangBlunt
PolymerasePhusion Plus DNA Polymerase
Quantity100 x 50 μL reactions
Reaction FormatSeparate components
Shipping ConditionRoom Temp or Wet Ice
Size (Final Product)20 kb or less
For Use With (Application)Hot-start PCR, High-fidelity PCR
GC-Rich PCR PerformanceHigh
Reaction Speed15 to 30 sec./kb
Unit SizeEach
Contents & Storage
• Phusion Plus DNA Polymerase (50 μL)
• 5X Phusion Plus Buffer (1.25 mL)
• 5X Phusion GC Enhancer (1.25 mL)

Store at –15°C to –25°C.

Frequently asked questions (FAQs)

What is the fidelity of the Phusion Plus DNA Polymerase (Cat. No. F630S)?

Please see "Conclusions" within the technical note for the Phusion Plus DNA Polymerase (Cat. No. F630S) (see link: https://assets.thermofisher.com/TFS-Assets/BID/Technical-Notes/measuring-error-rates-ngs-dna-polymerases-tech-note.pdf).

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Can I set up my reactions on ice if I am using the Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix (Cat. No. F631S)?

Reaction setup with Phusion Plus DNA Polymerase or Phusion Plus PCR Master Mix can be done either on ice or at room temperature.