본 제품은 액체 질소로 배송 및 보관되는 세포 관련 제품으로, 해당 제품 구매 시 세포 사용 가이드라인을 반드시 확인해주십시오. 세포사용가이드라인 다운로드
본 제품은 LMO 제품으로, 고객 분께서 LMO 신고 시스템을 통해 직접 수입 신고를 진행해주셔야 합니다. 자세히보기
Human OATP1B3 (G) SLC Transporter Cells
Gibco™
Human OATP1B3 (G) SLC Transporter Cells
TRANSiPORT Human OATP1B3 (G) SLC Transporter Cells are HEK293 cells that transiently overexpress the OATP1B3 solute carrier (SLC) transport protein.자세히 알아보기
Have Questions?
카탈로그 번호수량
GM1106G8–10M cells
카탈로그 번호 GM1106G
제품 가격(KRW)
832,000
온라인 행사
Ends: 30-Sep-2026
924,000
할인액 92,000 (10%)
Each
수량:
8–10M cells
제품 가격(KRW)
832,000
온라인 행사
Ends: 30-Sep-2026
924,000
할인액 92,000 (10%)
Each
TRANSiPORT Human OATP1B3 (G) SLC Transporter Cells are HEK293 cells that transiently overexpress the OATP1B3 solute carrier (SLC) transport protein. Solute carriers are a super-family of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes in and out of cells. The OATP1B3 SLC transporter protein is found in sinusoidal hepatocytes.
• Assess potential for transporter-mediated drug metabolism
• Easy to use format
• Obtain high quality results with a large signal-to-noise ratio
Choice of measurement system
Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.
Assay reliability
Mock HEK293 cells (Cat. No. GM1101G) that do not overexpress the SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of transport due to the presence of endogenous transporters or non-specific binding.
Rapid results
The convenient product format enables data generation in 2 days from thawing of the cells to final results.
Related products
GM1102G Human OATP1B1 (G) SLC Transporter Cells
GM1103G Human OAT1 (G) SLC Transporter Cells
GM1104G Human OAT3 (G) SLC Transporter Cells
GM1105G Human OCT2 (G) SLC Transporter Cells
GM1108G Human OCT1 (G) SLC Transporter Cells
GM1113G Human NTCP (G) SLC Transporter Cells
GM1114G Human MATE1 (G) SLC Transporter Cells
GM1115G Human MATE2K (G) SLC Transporter Cells
GM1101G Mock Cells
사양
셀 수8-10 x 106
제품 유형SLC Transporter Cell
수량8–10M cells
세포주293 (HEK)
Culture TypeMammalian Cell Culture
Human
Unit SizeEach
구성 및 보관
Store in liquid nitrogen vapor phase.
자주 묻는 질문(FAQ)
What are TRANSiPORT human solute carrier (SLC) transporter cells?
TRANSiPORT human SLC transporter cells are HEK293 cells that transiently overexpress a solute carrier (SLC) transporter protein. Solute carriers are one of two superfamilies of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes into and out of cells. Membrane transporters are found in hepatic cells, kidney proximal tubule cells, intestinal cells, and brain cells.
Find additional tips, troubleshooting help, and resources within our Drug Discovery & Development Support Center.
Is there a protocol available for the TRANSiPORT solute carrier (SLC) transporter cells?
Yes, the protocol is available here (https://www.genomembrane.com/information).
What should I use as a control with TRANSiPORT solute carrier (SLC) transporter cells?
Mock HEK293 cells (Cat. No. GM1001) that do not overexpress the SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of SLC transport due to the presence of endogenous transporters or non-specific binding.
With TRANSiPORT solute carrier (SLC) transporter cells, what should my choice of measurement be for SLC transporter activity?
Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.
How many cells can I expect in one vial of TRANSiPORT solute carrier (SLC) transporter cells?
One vial contains 8-12 million cells with high viability, designed to seed one 24-well plate for single use.