Human MATE2K (G) SLC Transporter Cells
Gibco™
Human MATE2K (G) SLC Transporter Cells
TRANSiPORT Human MATE2K (G) SLC Transporter Cells are HEK293 cells that transiently overexpress the MATE2K solute carrier (SLC) transport protein.Read more
Have Questions?
Catalog NumberQuantity
GM1115G8–10M cells
Catalog number GM1115G
Price (INR)
45,275.00
Online offer
Ends: 30-Sep-2026
82,318.00
Save 37,043.00 (45%)
Each
Quantity:
8–10M cells
Price (INR)
45,275.00
Online offer
Ends: 30-Sep-2026
82,318.00
Save 37,043.00 (45%)
Each
TRANSiPORT Human MATE2K (G) SLC Transporter Cells are HEK293 cells that transiently overexpress the MATE2K solute carrier (SLC) transport protein. Solute carriers are a super-family of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes in and out of cells. The MATE2K SLC transporter protein is found in kidney proximal tubule cells.
• Assess potential for transporter-mediated drug metabolism
• Easy to use format
• Obtain high quality results with a large signal-to-noise ratio
Choice of measurement system
Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.
Assay reliability
Mock HEK293 cells (Cat. No. GM1101G) that do not overexpress the SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of transport due to the presence of endogenous transporters or non-specific binding.
Rapid results
The convenient product format enables data generation in 2 days from thawing of the cells to final results.
Related products
GM1102G Human OATP1B1 (G) SLC Transporter Cells
GM1103G Human OAT1 (G) SLC Transporter Cells
GM1104G Human OAT3 (G) SLC Transporter Cells
GM1105G Human OCT2 (G) SLC Transporter Cells
GM1106G Human OATP1B3 (G) SLC Transporter Cells
GM1108G Human OCT1 (G) SLC Transporter Cells
GM1113G Human NTCP (G) SLC Transporter Cells
GM1114G Human MATE1 (G) SLC Transporter Cells
GM1101G Mock Cells
Specifications
No. of Cells8-10 x 106
Product TypeSLC Transporter Cell
Quantity8–10M cells
Cell Line293 (HEK)
Culture TypeMammalian Cell Culture
SpeciesHuman
Unit SizeEach
Contents & Storage
Store in liquid nitrogen vapor phase.
Frequently asked questions (FAQs)
What are TRANSiPORT human solute carrier (SLC) transporter cells?
TRANSiPORT human SLC transporter cells are HEK293 cells that transiently overexpress a solute carrier (SLC) transporter protein. Solute carriers are one of two superfamilies of membrane transporters that can affect pharmacokinetics and drug exposure by governing the transport of solutes into and out of cells. Membrane transporters are found in hepatic cells, kidney proximal tubule cells, intestinal cells, and brain cells.
Find additional tips, troubleshooting help, and resources within our Drug Discovery & Development Support Center.
Is there a protocol available for the TRANSiPORT solute carrier (SLC) transporter cells?
Yes, the protocol is available here (https://www.genomembrane.com/information).
What should I use as a control with TRANSiPORT solute carrier (SLC) transporter cells?
Mock HEK293 cells (Cat. No. GM1001) that do not overexpress the SLC transporter are available for use as a negative experimental control. Some compounds may demonstrate high background levels of SLC transport due to the presence of endogenous transporters or non-specific binding.
With TRANSiPORT solute carrier (SLC) transporter cells, what should my choice of measurement be for SLC transporter activity?
Cell-based assays can be performed using radioisotope-labeled compounds, fluorescence-labeled compounds, or non-labeled compounds. The amount of substrate transported into the cells can be measured directly using a liquid scintillation counter, fluorescence plate reader, or LC-MS/MS, thereby allowing direct evaluation of SLC transporter activity.
How many cells can I expect in one vial of TRANSiPORT solute carrier (SLC) transporter cells?
One vial contains 8-12 million cells with high viability, designed to seed one 24-well plate for single use.