The SKU IVGN2304 will be discontinued by December 31st 2026 or until the stock lasts. Alternative SKU EK0031 is available.
Product Image
Invitrogen™
Anza™ T4 PNK Kit
The Invitrogen™ Anza™ T4 PNK (polynucleotide kinase) Kit is used to perform 5’-phosphorylation of DNA and oligonucleotides. T4 PNK exhibitsRead more
Have Questions?
Catalog NumberQuantity
IVGN230450 reactions
Catalog number IVGN2304
Price (USD)
291.00
Each
Quantity:
50 reactions
Price (USD)
291.00
Each
The Invitrogen™ Anza™ T4 PNK (polynucleotide kinase) Kit is used to perform 5’-phosphorylation of DNA and oligonucleotides. T4 PNK exhibits 5’ polynucleotide kinase and 3’ phosphatase activity, catalyzing the transfer of the terminal phosphate of ATP to a 5’ hydroxyl group of a nucleic acid.
When preparing DNA for the ligation step in cloning, either the insert DNA or the vector DNA should contain a 5’ phosphate. Often PCR-amplified DNA is treated with T4 PNK to phosphorylate the 5’ end for subsequent cloning ligation.
Benefits:
• Fast 15-minute incubation at room temperature
• Phosphorylaton can be performed with DNA or oligonucleotides in water, TE, or 1X Anza buffers
The Anza T4 PNK Kit is part of the Anza™ Restriction Enzyme Cloning System, unifying the traditional cloning processes. Other Anza DNA Modifying Enzymes include the Anza T4 DNA Ligase Master Mix, Anza DNA Blunt End Kit, and Anza DNA End Repair Kit.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible BufferAnza™ 1X Buffer, Anza™ 10X T4 PNK Buffer
Product TypeT4 PNK Kit
Quantity50 reactions
Shipping ConditionApproved for shipment on Wet or Dry Ice
EnzymePolynucleotide Kinase
Product LineAnza
Unit SizeEach
Contents & Storage
50 reactions Anza T4 PNK Enzyme
0.5 mL Anza 10X T4 PNK Buffer
Store at -5 to -30°C.