Maxima SYBR Green/ROX qPCR Master Mix (2X)
Thermo Scientific™

Maxima SYBR Green/ROX qPCR Master Mix (2X)

**********************Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K0221200 reactions
K02221,000 Reactions
K02234,000 Reactions
Catalog number K0221
Price (MXN)
4,877.53
Each
No. of Reactions:
200 reactions
Price (MXN)
4,877.53
Each
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of Maxima SYBR Green qPCR Master Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific Maxima SYBR Green qPCR Master Mix is a ready-to-use solution optimized for qPCR and 2-step RT-qPCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG) (1). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most most real-time thermal cyclers (see Resources for a compatibility chart).

Highlights

• Specificity—Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
• Sensitivity—detects low copy number targets
• Wide linear range—accurate quantification across 9 orders of magnitude
• Reproducibility and convenience—ready-to-use 2x master mix

Applications

• Gene expression
• siRNA validation
• Genotyping
• Pathogen detection

Includes

Maxima SYBR Green/ROX qPCR Master Mix (2x) includes Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer supplemented with ROX passive reference dye. Supplied with nuclease-free water.

Related Products
Maxima SYBR Green/ROX qPCR Master Mix (2X)
Maxima SYBR Green/Fluorescein qPCR Master Mix (2X)
Maxima SYBR Green qPCR Master Mix (2X), with separate ROX vial
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
FormatTube
No. of Reactions200 reactions
PolymeraseTaq DNA Polymerase
Product LineMaxima
Product TypeReal Time PCR SYBR Master Mix
Quantity200 reactions
Sample TypeDNA, dsDNA
Sufficient For200 Reactions of 25 μL
Volume2 x 1.25 mL
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains:
• 2 x 1.25 mL Maxima SYBR Green/ROX qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase, dNTPs (also dUTP) and SYBR Green I in an optimized PCR buffer with ROX passive reference dye; sufficient for 200 x 25 μL reactions.
• 2 x 1.25 mL Nuclease-Free Water

Store at -20°C.

Frequently asked questions (FAQs)

Is the Maxima SYBR Green/ROX qPCR Master Mix (2X) (Cat. No. K0223, K0221, K0222) compatible with the ViiA 7 Real-Time PCR System (Cat. No. 4453542, 4453539)

No, the ViiA 7 Real-Time PCR System (Cat. No. 4453542, 4453539) is not compatible with the Maxima SYBR Green/ROX qPCR Master Mix (2X) (Cat. No. K0223).

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.