Thermo Scientific™

Maxima SYBR Green/Fluorescein qPCR Master Mix (2X)

Catalog number:  K0242

 Related applications:

Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of Maxima SYBR Green qPCR Master Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific Maxima SYBR Green qPCR Master Mix is a ready-to-use solution optimized for qPCR and 2-step RT-qPCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG) (1). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most most real-time thermal cyclers (see Resources for a compatibility chart).

Highlights

SpecificityMaxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
Sensitivity—detects low copy number targets
Wide linear range—accurate quantification across 9 orders of magnitude
Reproducibility and convenience—ready-to-use 2x master mix

Applications

• Gene expression
• siRNA validation
• Genotyping
• Pathogen detection

Includes

Maxima SYBR Green/Fluorescein qPCR Master Mix (2x) includes Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer supplemented with fluorescein passive reference dye. Supplied with nuclease-free water.

Related Products
Maxima SYBR Green/ROX qPCR Master Mix (2X)
Maxima SYBR Green/Fluorescein qPCR Master Mix (2X)
Maxima SYBR Green qPCR Master Mix (2X), with separate ROX vial
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: SYBR
For Use With (Equipment): BioRad MyiQ, BioRad iCycler iQ, BioRad iQ5
Reaction Speed: Standard
PCR Method: qPCR
Sample Type (General): DNA, dsDNA
Number of Reactions: 1,000 Reactions
Polymerase: Taq DNA Polymerase
Product Line: Maxima
Product Size: 1,000 reactions

Contents & storage

Contains:
• 10 x 1.25 mL Maxima SYBR Green/Fluorescein qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase, dNTPs (also dUTP) and SYBR Green I in an optimized PCR buffer with fluorescein passive reference dye; sufficient for 1000 x 25 µL reactions.
• 10 x 1.25 mL Nuclease-Free Water

Store at -20°C.

Documents

Manuals & protocols