Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • gamma-H2AX Antibodies

          Identifyn

          gamma-H2AX SRM Recombinant Rabbit Monoclonal Antibody (BLR014M), Identifyn™

          View all (17) gamma-H2AX antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite gamma-H2AX SRM Recombinant Rabbit Monoclonal Antibody (BLR014M), Identifyn™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (61)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 61

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          gamma-H2AX SRM Antibody (PA-RR000004) in ICC/IF

          IdentifynTM Detailed protocols can be found below under Documents & Downloads. Confocal Microscopy: HeLa Cells were grown on acid-etched 1.5H cover glass to ~50% confluency. DNA damage was induced via treatment with 2 µM Etoposide–spiked complete growth media for 24 hours, followed by -20°C methanol fixation and blocked with Thermo Fisher Fish Serum Blocker (Product # 37527X3). The primary antibody, Rabbit ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Western Blot (WB)
          gamma-H2AX SRM Antibody in Western Blot (WB)
          gamma-H2AX SRM Antibody in Western Blot (WB)
          gamma-H2AX SRM Antibody in Western Blot (WB)
          gamma-H2AX SRM Antibody in Western Blot (WB)
          gamma-H2AX SRM Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          gamma-H2AX SRM Recombinant Rabbit Monoclonal Antibody (BLR014M), Identifyn™

          Product Details

          PA-RR000004

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          -

          Immunocytochemistry (ICC/IF)

          1:200-1:500
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          ExpiCHO

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          BLR014M

          Immunogen

          Proprietary peptide specific to serine 139 phosphorylation.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 405 
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 555 
          • Alexa Fluor 594 
          • Alexa Fluor 647 
          • Alexa Fluor 680 
          • Alexa Fluor 750 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 10% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Identifyn™ Primary and Secondary antibodies are extensively validated in Immunofluorescence assays by traditional as well as high resolution acquisition like Widefield SIM, Confocal, Airyscan and STORM Microscopy.

          Recommended dilution for traditional microscopy:

          Widefield - 1:500

          Widefield SIM - 1:500

          Confocal - 1:500

          Confocal NIR - 1:500

          Recommended dilution for Super Resolution microscopy:

          SIM (Structured Illumination Microscopy) - 1:500

          Single Molecule STORM (Stochastic Optical Reconstruction Microscopy) - 1:200

          Detailed protocols can be found below under Methods & Protocols.

          Target Information

          H2AX is a variant of the H2A protein family and is a component of the histone octomer in nucleosomes. H2AX is phosphorylated by kinases such as ataxia telangiectasia mutated (ATM) and ATM-Rad3-related (ATR) in the PI3K pathway. Gamma-H2AX is the first step in recruiting and localizing DNA repair proteins. DSBs can be induced by mechanisms such as ionizing radiation or cytotoxic agents and subsequently, gamma-H2AX foci quickly form. These foci represent the DSBs in a 1:1 manner and can be used as a biomarker for damage. The visualization of gamma-H2AX by can allow the assessment of DNA damage, related DNA damage proteins and DNA repair. Gamma-H2AX has applications in the detection of genomic damage caused by cytotoxic chemical agents and environmental and physical damage.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: gamma H2A X; gamma H2A.X; gamma-H2A.X; gh2ax; H2A histone family member X; H2AFX; H2AX histone; histone H2A.x; unnamed protein product

          View more View less

          Gene Aliases: H2A.X; H2A/X; H2AFX

          View more View less

          UniProt ID: (Human) P16104

          View more View less

          Entrez Gene ID: (Human) 3014

          View more View less

          Function(s)
          DNA binding damaged DNA binding protein binding enzyme binding structural constituent of chromatin histone binding protein heterodimerization activity chromatin-protein adaptor activity
          Process(es)
          DNA damage checkpoint double-strand break repair nucleosome assembly cellular response to DNA damage stimulus response to ionizing radiation heterochromatin assembly positive regulation of DNA repair protein localization to site of double-strand break
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-jvpnf:80/100.66.74.151:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline