HyPerforma™ Single-Use Mixer DS 300, Unit with Drum Positioner and Adjustable Motor Head
Actual product may vary

HyPerforma™ Single-Use Mixer DS 300, Unit with Drum Positioner and Adjustable Motor Head

Have Questions?
카탈로그 번호 SUMDS03009003
제품 가격(KRW)
-
The Thermo Scientific™ HyPerforma™ DS 300 Single-Use Mixer (S.U.M.) is built upon the proven legacy design of the S.U.M; but optimized for lower-cost, high throughput applications. The smaller footprint and lighter weight of the HyPerforma S.U.M. DS 300 provide mobility and flexibility in mixing applications not currently available in the market.

Features:
• The docking station consists of a stainless steel base with locking casters, an adjustable handle, a tethered hand-held control device, an electronic vertical lift mechanism with integrated height indicator, motor mount and motor, three sizes of drive shafts, two adjustment tools—a spanner and a torque wrench. An adjustable-angle motor head and drum positioned are available as add-on options.
• Supporting hardware optimized for mixing, storage, transport and discharge in a closed system to help minimize the risk of cross-contamination.
• Top-drain support containers are available in 50, 100, 200 and 300 L sizes; with bottom-drain drums available in 50, 100 and 200 L sizes.
• Standard BPCs available for liquid/liquid and powder/liquid mixing.
• Optional dollies are available for all drum sizes.

Applications of the Thermo Scientific HyPerforma DS 300 S.U.M. include
• Hydration of powdered media and buffers.
• Sterile mixing of buffers, cell culture media, or other process fluids.
• Pooling of sterile liquid fractions.

Related products:
HyPerforma DS 300 Single-Use Mixers
• HyPerforma DS 300 Single-Use Mixer, Unit Standard
• HyPerforma DS 300 Single-Use Mixer, Unit with Drum Positioner
• HyPerforma DS 300 Single-Use Mixer, Unit with Adjustable Motor Head
• HyPerforma Mixer DS 300 Single-Use, Unit with Drum Positioner and Adjustable Motor Head

Drums for HyPerforma DS 300 Single-Use Mixers
• Plastic Drum, 50 L, with Conical Insert
• Plastic Drum, 100 L, with Conical Insert
• Plastic Drum, 200 L
• Plastic Drum, 300 L

BioProcess Containers (BPCs, bags and liners) for HyPerforma DS 300 Single-Use Mixers
• BioProcess Container (BPC), 50 L, Bottom drain, irradiated
• BioProcess Container (BPC), 100 L, Bottom drain, irradiated
• BioProcess Container (BPC), 500 L, Bottom drain, irradiated
• Bags and Liners, 50 L Liner - Bottom drain, non-irradiated
• Bags and Liners, 100 L Liner - Bottom drain, non-irradiated
• Bags and Liners, 200 L Liner - Bottom drain, non-irradiated

Dollys for for HyPerforma DS 300 Single-Use Mixers
• Dolly for 50 to 200 L Plastic Drums (single, polyethylene)
• Dolly for 50 to 200 L Plastic Drums (two, polyethylene)
• Dolly for 300 L Plastic Drum (stainless steel)
사양
Unit Size300 L