Bags and Liners for HyPerforma™ Single-Use Mixer DS 300, 200L Liner - Bottom drain, non-irradiated
Thermo Scientific™

Bags and Liners for HyPerforma™ Single-Use Mixer DS 300, 200L Liner - Bottom drain, non-irradiated

The 200L open-top, non-irradiated BioProcess Container (BPC) tank liner with bottom drain is composed of CX3-9 single web, multi-layer plastic film and compatible with Thermo Scientific™ HyPerforma Single-Use Mixer DS 300.
Have Questions?
Catalog NumberQuantity
SH3039903
also known as SH30399.03
200 L
Catalog number SH3039903
also known as SH30399.03
Price (USD)
-
Quantity:
200 L

The 200L open-top, non-irradiated BioProcess Container (BPC) tank liner with bottom drain is composed of CX3-9 single web, multi-layer plastic film and compatible with Thermo Scientific™ HyPerforma Single-Use Mixer DS 300. Implement industry-proven BPCs and liners in the overall process of shipping and storing your liquid-liquid and powder-liquid bioprocess mixing applications. BPCs and liners come in open- and closed-top design and are manufactured using our proprietary CX5-14 and CX3-9 film that easily fit in every plastic drum size.

Key Features of HyPerforma Single-Use Mixer DS 300 BPCs and liners:

  • All standard BPCs and liners are designed to fit 'out-of-the-box'
  • Available in volume options of 50, 100, and 200L
  • Applicable to open- and closed-top mixing applications
  • Powder-liquid and liquid-liquid mixing
  • World class CX5-14 and CX3-9 film
  • Adaptable to multiple upstream process applications in media and buffer prep

Required Accessories:

  • HyPerforma Single-Use Mixer DS 300 docking station
  • HyPerforma SIngle-Use Mixer DS 300 drums

Recommended for:

  • Powder-liquid or liquid-liquid mixing applications in single use
  • High-throughput mixing and storage
  • Additional configurations are available to suit your application requirements.
Specifications
DescriptionLiner - Bottom drain, non-irradiated
Film DescriptionCX3-9
Outer PackagingPackages placed in strong cardboard cartons with product and lot identification
Port DetailBottom Port: 0.5 in.
Product LineHyPerforma
Protective Inner PackagingPacked and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Quantity200 L
Sterilization MethodNon-Irradiated
Tubing Line 1Bottom Port: 1.27 x 1.91 x 122 cm (0.5 x 0.75 x 304.1 in.) Polycarbonate Quick Connect 12.7 mm (0.5 in.) insert (MPX) and cap
Capacity (Metric)200 L
Tubing StyleImpeller Sleeve with Quick Connect: C-Flex™ Tubing
Unit Size200 L