BioProcess Container (BPC), 100L, for HyPerforma™ Single-Use Mixer DS 300, Bottom drain, irradiated
Actual product may vary
BioProcess Container (BPC), 100L, for HyPerforma™ Single-Use Mixer DS 300, Bottom drain, irradiated
Have Questions?
카탈로그 번호 SH3094602
SH30946.02으로도 사용됨
제품 가격(KRW)
-
The 100L closed-top, irradiated BioProcess Container (BPC) with bottom drain is composed of CX3-9 single web, multi-layer plastic film and that fit inside drums for Thermo Scientific™ HyPerforma Single-Use Mixer DS 300. Implement industry-proven BPCs and liners in the overall process of shipping and storing your liquid-liquid and powder-liquid bioprocess mixing applications. BPCs and liners come in open- and closed-top design and are manufactured using our proprietary CX5-14 and CX3-9 film that easily fit in every plastic drum size.
Key features of HyPerforma Single-Use Mixer DS 300 BPCs and liners:
• All standard BPCs and liners are designed to fit “out-of-the-box“
• Available in volume options of 50, 100, and 200L
• Applicable to open- and closed-top mixing applications
• Powder-liquid and liquid-liquid mixing
• World class CX5-14 and CX3-9 film
• Adaptable to multiple upstream process applications in media and buffer prep
Required accessories:
• HyPerforma Single-Use Mixer DS 300 docking station
• HyPerforma SIngle-Use Mixer DS 300 drums
Recommended for:
• Powder-liquid or liquid-liquid mixing applications in single use
• High-throughput mixing and storage
• Additional configurations are available to suit your application requirements.
사양
Unit Size100 L