Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • TIGIT Antibodies

          Invitrogen

          TIGIT Monoclonal Antibody (MBSA43), Functional Grade, eBioscience™

          36 Published Figures
          38 References
          View all (101) TIGIT antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TIGIT Monoclonal Antibody (MBSA43), Functional Grade, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (36)
          TIGIT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          TIGIT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 37

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TIGIT Antibody (16-9500-38) in IHC (P)

          Immunohistochemical analysis of TIGIT was performed using formalin-fixed paraffin-embedded Human tonsil tissue sections. To expose the target protein, heat-induced epitope retrieval was performed on de-paraffinized sections using eBioscience™ IHC Antigen Retrieval Solution - Low pH (10X) (Product # 00-4955-58) diluted to 1X solution in water in a decloaking chamber at 110 degrees Celsius for 15 minutes. Fol... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TIGIT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TIGIT Antibody in Immunohistochemistry (IHC)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Flow Cytometry (Flow)
          TIGIT Antibody in Inhibition Assays (Inhibition)

          Product Details

          16-9500-38

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.25 µg/test
          View 27 publications 27 publications

          Functional Assay (Functional)

          Assay-Dependent
          View 3 publications 3 publications

          Inhibition Assays (Inhibition)

          -
          View 2 publications 2 publications

          Mass Spectrometry (MS)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), Functional Grade, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          MBSA43

          Conjugate

          Functional Grade Functional Grade Functional Grade
          • Alexa Fluor 488 
          • Alexa Fluor 647 
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • eFluor 450
          • FITC
          • NovaFluor Red 700
          • PE 
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          5 mg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2866362

          Product Specific Information

          Description: The MBSA43 monoclonal antibody reacts with human T cell immunoreceptor with Ig and ITIM domains (TIGIT), which is also known as VSTM3 or WUCAM. This 26 kDa protein is a member of the poliovirus receptor (PVR) family. The expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells, but it is not expressed on B cells or naive CD4+ T cells. TIGIT may be upregulated on naive CD4+ T cells upon activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK and T cells with antigen presenting cells, fibroblasts, and endothelial cells that express PVR and PVR-like proteins.

          MBSA43 is reported to crossreact with Rhesus macaque.

          Applications Reported: This MBSA43 antibody has been reported for use in flow cytometric analysis. In a functional assay, MBSA43 enhances IFN gamma secretion following TCR activation.

          Applications Tested: This MBSA43 antibody has been tested by flow cytometric analysis. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Storage and handling: Use in a sterile environment.

          Filtration: 0.2 µm post-manufacturing filtered.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Endotoxin Level: Less than 0.001 ng/µg antibody, as determined by LAL assay.

          Aggregation: Less than 10%, as determined by HPLC.

          Target Information

          TIGIT is a 26 kDa protein that belongs to the poliovirus receptor (PVR) family. It is expressed on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells, but not on B cells or naive CD4+ T cells. However, TIGIT can be upregulated on naive CD4+ T cells upon activation. TIGIT interacts with members of the PVR and PVR-like families, including CD155. This interaction mediates the engagement of NK and T cells with antigen-presenting cells, fibroblasts, and endothelial cells expressing PVR and PVR-like proteins. Binding of TIGIT to PVR leads to increased secretion of IL-10 and decreased secretion of IL-12B, promoting the generation of mature immunoregulatory dendritic cells and suppressing T cell activation. Through these interactions, TIGIT plays a significant role in modulating immune responses and maintaining immune homeostasis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: T-cell immunoreceptor with Ig and ITIM domains; unnamed protein product; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9, VSTM3; Washington University cell adhesion molecule

          View more View less

          Gene Aliases: TIGIT; VSIG9; VSTM3; WUCAM

          View more View less

          UniProt ID: (Human) Q495A1

          View more View less

          Entrez Gene ID: (Human) 201633

          View more View less

          Function(s)
          receptor binding protein binding signaling receptor activity identical protein binding
          Process(es)
          negative regulation of interleukin-12 production positive regulation of interleukin-10 production natural killer cell mediated cytotoxicity negative regulation of natural killer cell mediated cytotoxicity negative regulation of T cell activation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline