Power Blotter Pre-cut Membranes and Filters, PVDF mini
Invitrogen™
Power Blotter Pre-cut Membranes and Filters, PVDF mini
These Invitrogen Power Blotter Pre-cut Membranes and Filters are designed for transfer of proteins from mini-size gels using the PowerRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PB9220X33 x 20 stacks
PB922020 stacks
Catalog number PB9220X3
Price (EUR)
386,00
Each
Quantity:
3 x 20 stacks
Price (EUR)
386,00
Each
These Invitrogen Power Blotter Pre-cut Membranes and Filters are designed for transfer of proteins from mini-size gels using the Power Blotter System. The pre-cut membranes and filters include 20 sheets of polyvinylidene difluoride (PVDF) and 80 sheets of absorbent blotting paper (0.85 mm thickness). Power Blotter Pre-cut Membranes and Filters combined with Power Blotter 1-Step Transfer Buffer allow for high-efficiency semi-dry transfer of proteins in less than 10 minutes.
Features of PVDF membranes:
High-quality—PVDF transfer membranes manufactured especially for protein transfer and western blot applications
High-binding affinity—provides excellent protein binding (240 μg/cm2), blocks easily, and is compatible with most organic solvents, acids, and mild bases
Convenient—pre-cut PVDF sheets are ready to use for transfer and blotting experiments from standard precast or homemade mini-gels (8 cm x 8 cm); no need to cut your own membranes, minimizing damage and contamination
Validated—membrane and filter when combined provide the thickness and buffer capacity needed for optimal performance in the Power Blotter System
The PVDF membrane (0.45 μm pore size, low fluorescence) has higher binding capacity than nitrocellulose. The membrane is compatible with commonly used detection methods such as staining, immunodetection, and fluorescence. The proteins bind to the membrane through hydrophobic interactions. This membrane has lower background levels and increased sensitivity for fluorescent probing than regular PVDF or nitrocellulose membranes.
Learn more about the Power Blotter System ›
Related products
Power Blotter Pre-cut Membranes and Filters, PVDF, regular size
Power Blotter Pre-cut Membranes and Filters, nitrocellulose, mini
Power Blotter Pre-cut Membranes and Filters, nitrocellulose, regular size
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity3 x 20 stacks
Shipping ConditionRoom Temperature
Dimensions (LxW)7 x 8.4 cm
MaterialPVDF
Pore Size0.45 μm
Unit SizeEach
Contents & Storage
3 x 20 mini-size transfer stacks (8 x 8 cm) composed of 20 sheets of nitrocellulose and 80 sheets of filter paper (0.85 mm thickness)