Power Blotter 1-Step™ Transfer Buffer (5X)
Power Blotter 1-Step™ Transfer Buffer (5X)
Power Blotter 1-Step™ Transfer Buffer (5X)
Invitrogen™

Power Blotter 1-Step™ Transfer Buffer (5X)

Invitrogen Power Blotter 1-Step Transfer Buffer is designed for rapid semi-dry transfer of proteins from polyacrylamide gels (SDS-PAGE) to nitrocelluloseRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PB73001 L
PB7100250 mL
Catalog number PB7300
Price (MXN)
-
Quantity:
1 L
Invitrogen Power Blotter 1-Step Transfer Buffer is designed for rapid semi-dry transfer of proteins from polyacrylamide gels (SDS-PAGE) to nitrocellulose or PVDF membranes using the Power Blotter System. Transfer is compatible with commonly used detection methods such as staining, chemiluminescence, or fluorescence.

Features of Power Blotter 1-Step Transfer Buffer:
• Fast—high ionic strength formulation allows for 5- to 12-minute protein transfer when used with compatible fast semi-dry blotting systems
• Optimized—designed for seamless and reliable performance with the Power Blotter System
• Compatible—effective with other semi-dry transfer devices equipped with a suitable high-current power supply
• Economical—no need for special or costly consumables that are required by other fast semi-dry transfer devices

Learn more about the semi-dry transfer device: Power Blotter System

When used with the Power Blotter System, this buffer provides effective gel-to-membrane transfer of proteins in 5 to 12 minutes with efficiency that is similar to conventional western blotting techniques. Fast blotting methods require a high-current power supply, such as the Power Blotter System, and an optimized high ionic strength transfer buffer, such as Power Blotter 1-Step Transfer Buffer.

By increasing the current, excellent transfer efficiency can be achieved in much shorter time compared to conventional methods. Amperage is held at a constant rate based on the surface area of the transfer stack(s) (∼22–23 mA/cm2) and voltage is limited to 25 V.

Power Blotter 1-Step Transfer Buffer is also compatible with other protein semi-dry transfer devices, when they are paired with a suitable high-current power supply. Such devices provide constant high current (1.3 to 5.0 amps) to rapidly transfer proteins via the high ionic strength conditions supplied by the transfer buffer.

Related products
Power Blotter Pre-cut Membranes and Filters, nitrocellulose, regular size
Power Blotter Pre-cut Membranes and Filters, nitrocellulose, mini
Power Blotter Pre-cut Membranes and Filters, PVDF, regular size
Power Blotter Pre-cut Membranes and Filters, PVDF, mini

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Membrane CompatibilityNitrocellulose, PVDF
Mode of TransferSemi-Dry
Quantity1 L
Shipping ConditionRoom Temperature
For Use With (Application)Western Blot
Product Line1-Step
TypeTransfer Buffer
Unit SizeEach

Frequently asked questions (FAQs)

What is the difference between the Power Blotter System and the Power Blotter XL System?

The Power Blotter System (Cat. No. PB0012) comes with the Power Blotter Station (Cat. No. PB0010) plus Power Blotter Cassette (Cat. No. PB0002). The Power Blotter System can transfer one midi or up to two mini gels at the same time. The Power Blotter XL System (Cat. No. PB0013) on the other hand comes with the Power Blotter Station (Cat. No. PB0010) plus Power Blotter XL Cassette (Cat. No. PB0003). The Power Blotter XL System can transfer up to two midi or four mini gels at the same time.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Is it possible to do gel staining with the Power Blotter System/Power Blotter XL System?

Even though the Power Blotter System/Power Blotter XL System does not have the staining program, this can be introduced. Please see the user guide for our Pierce Mini/Midi Gel Power Staining Kit (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017799_22839_22840_powerstain_midi_minigel_UG.pdf) for how to create a Custom Staining Method.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What is the material used for the anode and cathode plates of the Power Blotter Cassette/Power Blotter Cassette XL?

The top (cathode) is stainless steel and the bottom (anode) is platinum-coated stainless steel.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

The central European or UK power cord for the Power Blotter System/Power Blotter XL System does not work in Switzerland. Do you offer a Swiss power cord?

Yes, you may purchase the Universal Cable Kit (Cat. No. 4377117).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

I have lost the power cord for the Power Blotter Station. Is it sold as a stand-alone item?

Here are the SKUs for the power cord. Please order the one appropriate for your region:

- Cat. No. 84858: Power Cord with C13 Connector, North America
- Cat. No. 84859: Power Cord with C13 Connector, Continental Europe and South Korea
- Cat. No. 84860: Power Cord with C13 Connector, United Kingdom and Singapore
- Cat. No. 84855: Chinese 10 Amp Cordset w/ C13 Connector
- Cat. No. 84856: Japanese 15 Amp Cordset w/ C13 Connector
- Cat. No. 84857: Australian 10 Amp Cordset w/ C13 Connector

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.